Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 1091687-1091936 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
C7orf50
Entrez Gene ID
84310
Gene Name
chromosome 7 open reading frame 50
Gene Aliases
YCR016W
UniGene
Hs.653258
Gene Chromosome Location
Chr.7: 978485-1138291 on Build GRCh38
Transcripts
NM_001134395.1 NM_001134396.1 NM_001318252.1 NM_032350.5 XM_011515581.2 XM_011515582.2 XM_011515583.2 XM_011515584.2 XM_017012720.1 XM_017012721.1 XM_017012722.1 NM_001039966.1 NM_001098201.1 NM_001505.2

Gene Symbol
GPER1
Entrez Gene ID
2852
Gene Name
G protein-coupled estrogen receptor 1
Gene Aliases
CEPR, CMKRL2, DRY12, FEG-1, GPCR-Br, GPER, GPR30, LERGU, LERGU2, LyGPR, mER
UniGene
Hs.20961
Gene Chromosome Location
Chr.7: 1086807-1093815 on Build GRCh38
Transcripts
NM_001134395.1 NM_001134396.1 NM_001318252.1 NM_032350.5 XM_011515581.2 XM_011515582.2 XM_011515583.2 XM_011515584.2 XM_017012720.1 XM_017012721.1 XM_017012722.1 NM_001039966.1 NM_001098201.1 NM_001505.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3674803 c.84C>A p.S28S Chr.7: 1091812-1091812 on Build GRCh38
7328677 c.90G>A p.E30E Chr.7: 1091818-1091818 on Build GRCh38
1084157 c.186G>A p.S62S Chr.7: 1091914-1091914 on Build GRCh38
7269721 c.3G>T p.M1I Chr.7: 1091731-1091731 on Build GRCh38
4687791 c.81C>A p.T27T Chr.7: 1091809-1091809 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs769072422 Chr.7: 1091790-1091790 on Build GRCh38
rs760554923 Chr.7: 1091739-1091739 on Build GRCh38
rs771081864 Chr.7: 1091888-1091888 on Build GRCh38
rs201954771 Chr.7: 1091816-1091816 on Build GRCh38
rs773117474 Chr.7: 1091818-1091818 on Build GRCh38
rs11544331 Chr.7: 1091775-1091775 on Build GRCh38
rs758280016 Chr.7: 1091761-1091761 on Build GRCh38
rs953609494 Chr.7: 1091877-1091877 on Build GRCh38
rs760777642 Chr.7: 1091760-1091760 on Build GRCh38
rs988700324 Chr.7: 1091800-1091800 on Build GRCh38
rs985757418 Chr.7: 1091711-1091711 on Build GRCh38
rs199734759 Chr.7: 1091815-1091815 on Build GRCh38
rs767235083 Chr.7: 1091911-1091911 on Build GRCh38
rs150332534 Chr.7: 1091878-1091878 on Build GRCh38
rs769010336 Chr.7: 1091753-1091753 on Build GRCh38
rs767361838 Chr.7: 1091835-1091835 on Build GRCh38
rs769056476 Chr.7: 1091759-1091760 on Build GRCh38
rs761757323 Chr.7: 1091908-1091908 on Build GRCh38
rs1018989115 Chr.7: 1091889-1091889 on Build GRCh38
rs61735233 Chr.7: 1091756-1091756 on Build GRCh38
rs201822599 Chr.7: 1091755-1091755 on Build GRCh38
rs750220402 Chr.7: 1091914-1091914 on Build GRCh38
rs769930139 Chr.7: 1091899-1091899 on Build GRCh38
rs755729056 Chr.7: 1091838-1091838 on Build GRCh38
rs752616345 Chr.7: 1091730-1091730 on Build GRCh38
rs774857621 Chr.7: 1091754-1091754 on Build GRCh38
rs926969843 Chr.7: 1091849-1091849 on Build GRCh38
rs144934685 Chr.7: 1091893-1091893 on Build GRCh38
rs763431580 Chr.7: 1091751-1091751 on Build GRCh38
rs772551045 Chr.7: 1091872-1091872 on Build GRCh38
rs767636021 Chr.7: 1091721-1091721 on Build GRCh38
rs772601689 Chr.7: 1091801-1091801 on Build GRCh38
rs368800854 Chr.7: 1091717-1091717 on Build GRCh38
rs370224401 Chr.7: 1091793-1091793 on Build GRCh38
rs3802141 Chr.7: 1091720-1091720 on Build GRCh38
rs200264603 Chr.7: 1091718-1091718 on Build GRCh38
rs571316739 Chr.7: 1091874-1091874 on Build GRCh38
rs140084051 Chr.7: 1091848-1091848 on Build GRCh38
rs776064895 Chr.7: 1091750-1091750 on Build GRCh38
rs541413145 Chr.7: 1091764-1091764 on Build GRCh38
rs748731423 Chr.7: 1091862-1091862 on Build GRCh38
rs149599681 Chr.7: 1091733-1091733 on Build GRCh38
rs774261850 Chr.7: 1091907-1091907 on Build GRCh38
rs780791783 Chr.7: 1091782-1091782 on Build GRCh38
rs779527973 Chr.7: 1091850-1091850 on Build GRCh38
rs369178618 Chr.7: 1091785-1091785 on Build GRCh38
rs146678860 Chr.7: 1091794-1091794 on Build GRCh38
rs981115047 Chr.7: 1091844-1091844 on Build GRCh38
rs757042408 Chr.7: 1091833-1091833 on Build GRCh38
rs763997927 Chr.7: 1091824-1091824 on Build GRCh38
rs148463493 Chr.7: 1091749-1091749 on Build GRCh38
rs748768043 Chr.7: 1091847-1091847 on Build GRCh38
rs746604818 Chr.7: 1091734-1091734 on Build GRCh38
rs201711034 Chr.7: 1091828-1091828 on Build GRCh38
rs750559019 Chr.7: 1091726-1091726 on Build GRCh38
rs778884454 Chr.7: 1091729-1091729 on Build GRCh38
rs936999827 Chr.7: 1091873-1091873 on Build GRCh38
rs779447108 Chr.7: 1091784-1091784 on Build GRCh38
rs759997674 Chr.7: 1091715-1091715 on Build GRCh38
rs117290655 Chr.7: 1091742-1091742 on Build GRCh38
rs376137544 Chr.7: 1091783-1091783 on Build GRCh38
rs759504275 Chr.7: 1091894-1091894 on Build GRCh38
rs115028942 Chr.7: 1091776-1091776 on Build GRCh38
rs770603037 Chr.7: 1091735-1091735 on Build GRCh38
rs754818831 Chr.7: 1091727-1091727 on Build GRCh38
rs779573563 Chr.7: 1091841-1091841 on Build GRCh38
rs199519155 Chr.7: 1091836-1091836 on Build GRCh38
rs760862902 Chr.7: 1091813-1091813 on Build GRCh38
rs576633880 Chr.7: 1091747-1091747 on Build GRCh38
rs34497267 Chr.7: 1091758-1091758 on Build GRCh38
rs773563688 Chr.7: 1091812-1091812 on Build GRCh38

View Full Product Details