Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.15: 88639284-88639529 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ISG20
Entrez Gene ID
3669
Gene Name
interferon stimulated exonuclease gene 20
Gene Aliases
CD25, HEM45
UniGene
Hs.459265
Gene Chromosome Location
Chr.15: 88635614-88656344 on Build GRCh38
Transcripts
NM_001303233.1 NM_001303234.1 NM_001303235.1 NM_001303236.1 NM_001303237.1 NM_002201.5 NR_130134.1 XM_005254899.2 XM_006720488.3 XM_017022147.1 XM_017022148.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
2014613 c.31G>A p.D11N Chr.15: 88639397-88639397 on Build GRCh38
3932188 c.79C>T p.R27C Chr.15: 88639445-88639445 on Build GRCh38
5653177 c.87C>G p.S29R Chr.15: 88639453-88639453 on Build GRCh38
391122 c.48_55delGCTGGGGC p.L17fs*27 Chr.15: 88639414-88639421 on Build GRCh38
7372496 c.64G>T p.E22* Chr.15: 88639430-88639430 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs771740851 Chr.15: 88639470-88639470 on Build GRCh38
rs768178202 Chr.15: 88639348-88639348 on Build GRCh38
rs771607779 Chr.15: 88639481-88639481 on Build GRCh38
rs759880476 Chr.15: 88639487-88639487 on Build GRCh38
rs547664272 Chr.15: 88639447-88639447 on Build GRCh38
rs202155196 Chr.15: 88639380-88639380 on Build GRCh38
rs35008759 Chr.15: 88639471-88639471 on Build GRCh38
rs865959092 Chr.15: 88639463-88639463 on Build GRCh38
rs747496926 Chr.15: 88639479-88639479 on Build GRCh38
rs535341571 Chr.15: 88639385-88639385 on Build GRCh38
rs774668729 Chr.15: 88639346-88639346 on Build GRCh38
rs144037981 Chr.15: 88639456-88639456 on Build GRCh38
rs904198974 Chr.15: 88639446-88639446 on Build GRCh38
rs911681615 Chr.15: 88639397-88639397 on Build GRCh38
rs762977706 Chr.15: 88639497-88639497 on Build GRCh38
rs377690718 Chr.15: 88639361-88639361 on Build GRCh38
rs867689427 Chr.15: 88639409-88639409 on Build GRCh38
rs754205235 Chr.15: 88639434-88639434 on Build GRCh38
rs1042739484 Chr.15: 88639312-88639312 on Build GRCh38
rs558554632 Chr.15: 88639461-88639461 on Build GRCh38
rs773881244 Chr.15: 88639402-88639402 on Build GRCh38
rs768350346 Chr.15: 88639468-88639468 on Build GRCh38
rs533916132 Chr.15: 88639457-88639457 on Build GRCh38
rs752810937 Chr.15: 88639445-88639445 on Build GRCh38
rs3200942 Chr.15: 88639365-88639365 on Build GRCh38
rs936674774 Chr.15: 88639475-88639475 on Build GRCh38
rs757187331 Chr.15: 88639377-88639377 on Build GRCh38
rs565872324 Chr.15: 88639453-88639453 on Build GRCh38
rs763950539 Chr.15: 88639377-88639378 on Build GRCh38
rs781166379 Chr.15: 88639379-88639379 on Build GRCh38
rs781049623 Chr.15: 88639311-88639311 on Build GRCh38
rs773861899 Chr.15: 88639420-88639420 on Build GRCh38
rs747603111 Chr.15: 88639469-88639469 on Build GRCh38
rs754123126 Chr.15: 88639362-88639362 on Build GRCh38
rs552242478 Chr.15: 88639319-88639319 on Build GRCh38
rs771750914 Chr.15: 88639435-88639435 on Build GRCh38
rs779530925 Chr.15: 88639383-88639383 on Build GRCh38
rs752763398 Chr.15: 88639369-88639369 on Build GRCh38
rs768239945 Chr.15: 88639343-88639343 on Build GRCh38
rs970036133 Chr.15: 88639321-88639324 on Build GRCh38
rs775133980 Chr.15: 88639324-88639324 on Build GRCh38
rs61733732 Chr.15: 88639462-88639462 on Build GRCh38
rs779029611 Chr.15: 88639367-88639367 on Build GRCh38
rs138335170 Chr.15: 88639322-88639322 on Build GRCh38
rs761178343 Chr.15: 88639357-88639357 on Build GRCh38
rs903805874 Chr.15: 88639472-88639472 on Build GRCh38
rs777053263 Chr.15: 88639483-88639483 on Build GRCh38
rs761294561 Chr.15: 88639403-88639403 on Build GRCh38
rs750810463 Chr.15: 88639356-88639356 on Build GRCh38
rs761082582 Chr.15: 88639427-88639427 on Build GRCh38
rs765448034 Chr.15: 88639439-88639439 on Build GRCh38
rs775583025 Chr.15: 88639496-88639496 on Build GRCh38
rs1025149663 Chr.15: 88639352-88639352 on Build GRCh38
rs751684388 Chr.15: 88639499-88639499 on Build GRCh38
rs762300315 Chr.15: 88639339-88639339 on Build GRCh38
rs142387462 Chr.15: 88639495-88639495 on Build GRCh38
rs770868397 Chr.15: 88639502-88639502 on Build GRCh38
rs903879496 Chr.15: 88639320-88639320 on Build GRCh38
rs1017593700 Chr.15: 88639327-88639327 on Build GRCh38
rs139144975 Chr.15: 88639410-88639410 on Build GRCh38
rs916535224 Chr.15: 88639425-88639425 on Build GRCh38

View Full Product Details