Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 67090023-67090284 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
MAST4
Entrez Gene ID
375449
Gene Name
microtubule associated serine/threonine kinase family member 4
Gene Aliases
-
UniGene
Hs.595458
Gene Chromosome Location
Chr.5: 66596348-67169595 on Build GRCh38
Transcripts
NM_001164664.1 NM_001290226.1 NM_001290227.1 NM_001297651.1 NM_015183.2 XM_006714606.3 XM_006714610.2 XM_011543382.2 XM_011543384.2 XM_011543385.2 XM_011543386.2 XM_017009447.1 XM_017009448.1 XM_017009449.1 XM_017009450.1 XM_017009451.1 XM_017009452.1 XM_017009453.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6311929 c.259G>T p.A87S Chr.5: 67090203-67090203 on Build GRCh38
3776806 c.240A>G p.S80S Chr.5: 67090220-67090220 on Build GRCh38
6895857 c.246C>T p.A82A Chr.5: 67090211-67090211 on Build GRCh38
4348515 c.265A>G p.R89G Chr.5: 67090230-67090230 on Build GRCh38
5429649 c.830G>A p.R277Q Chr.5: 67090228-67090228 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs775846433 Chr.5: 67090200-67090200 on Build GRCh38
rs759741352 Chr.5: 67090158-67090158 on Build GRCh38
rs953720854 Chr.5: 67090086-67090086 on Build GRCh38
rs771095135 Chr.5: 67090141-67090141 on Build GRCh38
rs977815695 Chr.5: 67090159-67090159 on Build GRCh38
rs773537698 Chr.5: 67090129-67090129 on Build GRCh38
rs371008871 Chr.5: 67090220-67090220 on Build GRCh38
rs1029265385 Chr.5: 67090077-67090077 on Build GRCh38
rs767358421 Chr.5: 67090218-67090218 on Build GRCh38
rs772492131 Chr.5: 67090115-67090115 on Build GRCh38
rs968241698 Chr.5: 67090143-67090143 on Build GRCh38
rs776821341 Chr.5: 67090152-67090152 on Build GRCh38
rs771579108 Chr.5: 67090140-67090141 on Build GRCh38
rs747577198 Chr.5: 67090131-67090131 on Build GRCh38
rs762581118 Chr.5: 67090063-67090063 on Build GRCh38
rs765544422 Chr.5: 67090191-67090191 on Build GRCh38
rs368492919 Chr.5: 67090114-67090114 on Build GRCh38
rs75626107 Chr.5: 67090102-67090102 on Build GRCh38
rs761445923 Chr.5: 67090202-67090202 on Build GRCh38
rs372195966 Chr.5: 67090242-67090242 on Build GRCh38
rs572011054 Chr.5: 67090111-67090111 on Build GRCh38
rs766339253 Chr.5: 67090244-67090244 on Build GRCh38

View Full Product Details