Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 43628053-43628306 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
NNT
Entrez Gene ID
23530
Gene Name
nicotinamide nucleotide transhydrogenase
Gene Aliases
GCCD4
UniGene
Hs.482043
Gene Chromosome Location
Chr.5: 43602574-43705566 on Build GRCh38
Transcripts
NM_012343.3 NM_182977.2 XM_005248274.4 XM_005248275.4 XM_006714461.3 XM_011514001.2 XM_011514002.2 XM_017009293.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6301955 c.822G>C p.L274F Chr.5: 43628245-43628245 on Build GRCh38
3920127 c.798C>T p.F266F Chr.5: 43628221-43628221 on Build GRCh38
3616754 c.819C>T p.P273P Chr.5: 43628242-43628242 on Build GRCh38
5785233 c.806T>C p.L269P Chr.5: 43628229-43628229 on Build GRCh38
3941332 c.830A>G p.D277G Chr.5: 43628253-43628253 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs757697012 Chr.5: 43628254-43628254 on Build GRCh38
rs746831453 Chr.5: 43628189-43628189 on Build GRCh38
rs763475950 Chr.5: 43628234-43628234 on Build GRCh38
rs545923662 Chr.5: 43628170-43628170 on Build GRCh38
rs754526914 Chr.5: 43628160-43628160 on Build GRCh38
rs373124357 Chr.5: 43628211-43628211 on Build GRCh38
rs775472540 Chr.5: 43628215-43628215 on Build GRCh38
rs991431174 Chr.5: 43628145-43628145 on Build GRCh38
rs981895670 Chr.5: 43628163-43628163 on Build GRCh38
rs200099620 Chr.5: 43628153-43628153 on Build GRCh38
rs919359469 Chr.5: 43628181-43628181 on Build GRCh38
rs374128623 Chr.5: 43628149-43628149 on Build GRCh38
rs778457611 Chr.5: 43628274-43628274 on Build GRCh38
rs147924268 Chr.5: 43628213-43628213 on Build GRCh38
rs957253582 Chr.5: 43628121-43628121 on Build GRCh38
rs776843409 Chr.5: 43628196-43628196 on Build GRCh38
rs771946402 Chr.5: 43628173-43628173 on Build GRCh38
rs764529261 Chr.5: 43628239-43628239 on Build GRCh38
rs141799831 Chr.5: 43628249-43628249 on Build GRCh38
rs763634624 Chr.5: 43628190-43628190 on Build GRCh38
rs201853415 Chr.5: 43628180-43628180 on Build GRCh38
rs535321543 Chr.5: 43628088-43628088 on Build GRCh38
rs778348769 Chr.5: 43628161-43628161 on Build GRCh38
rs1032905681 Chr.5: 43628107-43628107 on Build GRCh38
rs748098882 Chr.5: 43628164-43628164 on Build GRCh38
rs147111520 Chr.5: 43628260-43628260 on Build GRCh38
rs747645913 Chr.5: 43628277-43628277 on Build GRCh38
rs998544547 Chr.5: 43628092-43628092 on Build GRCh38

View Full Product Details