Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 116599984-116600249 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PNLIPRP1
Entrez Gene ID
5407
Gene Name
pancreatic lipase related protein 1
Gene Aliases
PLRP1
UniGene
Hs.73923
Gene Chromosome Location
Chr.10: 116590415-116609175 on Build GRCh38
Transcripts
NM_001303135.1 NM_006229.3 XM_011539868.1 XM_011539869.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7384795 c.908G>T p.C303F Chr.10: 116600140-116600140 on Build GRCh38
4822228 c.821G>T p.R274L Chr.10: 116600053-116600053 on Build GRCh38
5969674 c.838A>T p.N280Y Chr.10: 116600070-116600070 on Build GRCh38
2054754 c.908G>T p.C303F Chr.10: 116600140-116600140 on Build GRCh38
365620 c.867G>A p.L289L Chr.10: 116600099-116600099 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs781855279 Chr.10: 116600186-116600186 on Build GRCh38
rs199855159 Chr.10: 116600014-116600014 on Build GRCh38
rs781995141 Chr.10: 116600115-116600115 on Build GRCh38
rs199754358 Chr.10: 116600052-116600052 on Build GRCh38
rs527543102 Chr.10: 116600173-116600173 on Build GRCh38
rs782651844 Chr.10: 116600046-116600046 on Build GRCh38
rs782107666 Chr.10: 116600117-116600117 on Build GRCh38
rs372862202 Chr.10: 116600178-116600178 on Build GRCh38
rs782470470 Chr.10: 116600193-116600193 on Build GRCh38
rs17848304 Chr.10: 116600185-116600185 on Build GRCh38
rs782402071 Chr.10: 116600109-116600109 on Build GRCh38
rs150148236 Chr.10: 116600049-116600049 on Build GRCh38
rs781919453 Chr.10: 116600104-116600104 on Build GRCh38
rs782422123 Chr.10: 116600208-116600208 on Build GRCh38
rs782581906 Chr.10: 116600194-116600194 on Build GRCh38
rs782753215 Chr.10: 116600152-116600152 on Build GRCh38
rs959649900 Chr.10: 116600118-116600118 on Build GRCh38
rs782611633 Chr.10: 116600037-116600037 on Build GRCh38
rs782213653 Chr.10: 116600097-116600097 on Build GRCh38
rs782387571 Chr.10: 116600056-116600056 on Build GRCh38
rs782329170 Chr.10: 116600099-116600099 on Build GRCh38
rs782706605 Chr.10: 116600168-116600168 on Build GRCh38
rs782075346 Chr.10: 116600160-116600160 on Build GRCh38
rs782154349 Chr.10: 116600108-116600108 on Build GRCh38
rs782278062 Chr.10: 116600053-116600053 on Build GRCh38
rs782524165 Chr.10: 116600024-116600024 on Build GRCh38
rs782204334 Chr.10: 116600040-116600040 on Build GRCh38
rs782659758 Chr.10: 116600214-116600214 on Build GRCh38
rs267602380 Chr.10: 116600136-116600136 on Build GRCh38
rs138586853 Chr.10: 116600122-116600122 on Build GRCh38
rs782369072 Chr.10: 116600171-116600171 on Build GRCh38
rs3816355 Chr.10: 116600036-116600036 on Build GRCh38
rs56373266 Chr.10: 116600005-116600005 on Build GRCh38
rs781897399 Chr.10: 116600012-116600012 on Build GRCh38
rs765840584 Chr.10: 116600055-116600055 on Build GRCh38
rs782536087 Chr.10: 116600071-116600072 on Build GRCh38
rs376188042 Chr.10: 116600027-116600027 on Build GRCh38
rs782638666 Chr.10: 116600182-116600182 on Build GRCh38
rs781969680 Chr.10: 116600088-116600088 on Build GRCh38
rs782775789 Chr.10: 116600025-116600025 on Build GRCh38

View Full Product Details