Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 203955417-203955678 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ICOS
Entrez Gene ID
29851
Gene Name
inducible T-cell costimulator
Gene Aliases
AILIM, CD278, CVID1
UniGene
Hs.56247
Gene Chromosome Location
Chr.2: 203936748-203961577 on Build GRCh38
Transcripts
NM_012092.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4455047 c.64A>T p.I22F Chr.2: 203955641-203955641 on Build GRCh38
442155 c.85G>A p.E29K Chr.2: 203955662-203955662 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs370808791 Chr.2: 203955610-203955610 on Build GRCh38
rs55877101 Chr.2: 203955482-203955482 on Build GRCh38
rs758295536 Chr.2: 203955623-203955623 on Build GRCh38
rs1015881666 Chr.2: 203955638-203955638 on Build GRCh38
rs748491462 Chr.2: 203955605-203955605 on Build GRCh38
rs1019533261 Chr.2: 203955516-203955516 on Build GRCh38
rs958469813 Chr.2: 203955604-203955604 on Build GRCh38
rs764098873 Chr.2: 203955633-203955633 on Build GRCh38
rs997089111 Chr.2: 203955574-203955574 on Build GRCh38
rs778800567 Chr.2: 203955471-203955471 on Build GRCh38
rs775317381 Chr.2: 203955626-203955626 on Build GRCh38
rs1028572358 Chr.2: 203955584-203955584 on Build GRCh38
rs769294495 Chr.2: 203955624-203955624 on Build GRCh38
rs1027508589 Chr.2: 203955444-203955444 on Build GRCh38
rs541625712 Chr.2: 203955457-203955457 on Build GRCh38
rs776344551 Chr.2: 203955615-203955615 on Build GRCh38
rs779201161 Chr.2: 203955602-203955602 on Build GRCh38
rs965608370 Chr.2: 203955518-203955518 on Build GRCh38
rs745431176 Chr.2: 203955620-203955620 on Build GRCh38
rs900363907 Chr.2: 203955497-203955497 on Build GRCh38
rs1039356990 Chr.2: 203955476-203955476 on Build GRCh38
rs56326865 Chr.2: 203955498-203955498 on Build GRCh38
rs762930448 Chr.2: 203955630-203955630 on Build GRCh38
rs1051522004 Chr.2: 203955567-203955567 on Build GRCh38
rs547348342 Chr.2: 203955576-203955576 on Build GRCh38

View Full Product Details