Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 26746278-26746535 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PDSS1
Entrez Gene ID
23590
Gene Name
prenyl (decaprenyl) diphosphate synthase, subunit 1
Gene Aliases
COQ1, COQ10D2, DPS, SPS, TPRT, TPT, TPT 1, hDPS1
UniGene
Hs.558468
Gene Chromosome Location
Chr.10: 26697052-26746798 on Build GRCh38
Transcripts
NM_001321978.1 NM_001321979.1 NM_014317.4 XM_011519437.2 XM_017016011.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4655497 c.1202A>G p.D401G Chr.10: 26746427-26746427 on Build GRCh38
267160 c.1138G>A p.A380T Chr.10: 26746363-26746363 on Build GRCh38
5461835 c.1156G>A p.E386K Chr.10: 26746381-26746381 on Build GRCh38
5836719 c.1159G>A p.A387T Chr.10: 26746384-26746384 on Build GRCh38
1603356 c.1108-9A>G p.? Chr.10: 26746324-26746324 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs749681029 Chr.10: 26746501-26746501 on Build GRCh38
rs751585593 Chr.10: 26746468-26746468 on Build GRCh38
rs138571988 Chr.10: 26746472-26746472 on Build GRCh38
rs750246191 Chr.10: 26746435-26746435 on Build GRCh38
rs768647933 Chr.10: 26746500-26746501 on Build GRCh38
rs115180525 Chr.10: 26746476-26746476 on Build GRCh38
rs199603576 Chr.10: 26746419-26746419 on Build GRCh38
rs115687408 Chr.10: 26746382-26746382 on Build GRCh38
rs750436121 Chr.10: 26746325-26746325 on Build GRCh38
rs763824348 Chr.10: 26746310-26746310 on Build GRCh38
rs779407167 Chr.10: 26746323-26746324 on Build GRCh38
rs975467562 Chr.10: 26746408-26746408 on Build GRCh38
rs758061713 Chr.10: 26746355-26746355 on Build GRCh38
rs757187875 Chr.10: 26746322-26746322 on Build GRCh38
rs771476505 Chr.10: 26746393-26746393 on Build GRCh38
rs747125868 Chr.10: 26746345-26746345 on Build GRCh38
rs771227381 Chr.10: 26746498-26746499 on Build GRCh38
rs760294360 Chr.10: 26746405-26746405 on Build GRCh38
rs763649433 Chr.10: 26746417-26746417 on Build GRCh38
rs999754695 Chr.10: 26746460-26746460 on Build GRCh38
rs777891711 Chr.10: 26746472-26746473 on Build GRCh38
rs989486151 Chr.10: 26746329-26746329 on Build GRCh38
rs761681069 Chr.10: 26746429-26746429 on Build GRCh38
rs766397686 Chr.10: 26746467-26746467 on Build GRCh38
rs758471892 Chr.10: 26746463-26746464 on Build GRCh38
rs1057519354 Chr.10: 26746333-26746333 on Build GRCh38
rs778962956 Chr.10: 26746324-26746324 on Build GRCh38
rs201068515 Chr.10: 26746368-26746368 on Build GRCh38
rs770190130 Chr.10: 26746373-26746373 on Build GRCh38
rs371752451 Chr.10: 26746431-26746431 on Build GRCh38
rs749721462 Chr.10: 26746387-26746387 on Build GRCh38
rs916340943 Chr.10: 26746484-26746484 on Build GRCh38
rs779930386 Chr.10: 26746334-26746334 on Build GRCh38
rs922702614 Chr.10: 26746414-26746414 on Build GRCh38
rs574624767 Chr.10: 26746440-26746440 on Build GRCh38
rs747179524 Chr.10: 26746481-26746485 on Build GRCh38
rs776310500 Chr.10: 26746477-26746477 on Build GRCh38
rs748477018 Chr.10: 26746363-26746363 on Build GRCh38
rs202175213 Chr.10: 26746330-26746330 on Build GRCh38
rs367942639 Chr.10: 26746326-26746326 on Build GRCh38
rs755121257 Chr.10: 26746470-26746470 on Build GRCh38
rs753674730 Chr.10: 26746314-26746314 on Build GRCh38
rs781457293 Chr.10: 26746359-26746359 on Build GRCh38
rs112661893 Chr.10: 26746411-26746411 on Build GRCh38
rs115460991 Chr.10: 26746488-26746488 on Build GRCh38
rs748160237 Chr.10: 26746334-26746335 on Build GRCh38
rs773653686 Chr.10: 26746496-26746497 on Build GRCh38
rs114003104 Chr.10: 26746304-26746304 on Build GRCh38
rs754262066 Chr.10: 26746322-26746323 on Build GRCh38
rs778047720 Chr.10: 26746489-26746489 on Build GRCh38
rs879058849 Chr.10: 26746301-26746301 on Build GRCh38
rs774809733 Chr.10: 26746401-26746401 on Build GRCh38

View Full Product Details