Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 45061231-45061488 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
NMT1
Entrez Gene ID
4836
Gene Name
N-myristoyltransferase 1
Gene Aliases
NMT
UniGene
Hs.532790
Gene Chromosome Location
Chr.17: 45061291-45109016 on Build GRCh38
Transcripts
NM_021079.4 XM_011524858.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4804738 c.101A>T p.E34V Chr.17: 45061430-45061430 on Build GRCh38
4801845 c.131+1G>T p.? Chr.17: 45061461-45061461 on Build GRCh38
1302907 c.88T>C p.C30R Chr.17: 45061417-45061417 on Build GRCh38
4824316 c.10G>A p.E4K Chr.17: 45061339-45061339 on Build GRCh38
3191829 c.100G>C p.E34Q Chr.17: 45061429-45061429 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs760309598 Chr.17: 45061340-45061340 on Build GRCh38
rs768521050 Chr.17: 45061319-45061319 on Build GRCh38
rs764406772 Chr.17: 45061352-45061352 on Build GRCh38
rs1022554088 Chr.17: 45061349-45061349 on Build GRCh38
rs564172691 Chr.17: 45061342-45061342 on Build GRCh38
rs373569442 Chr.17: 45061299-45061299 on Build GRCh38
rs768076232 Chr.17: 45061431-45061431 on Build GRCh38
rs749106795 Chr.17: 45061317-45061317 on Build GRCh38
rs267604914 Chr.17: 45061345-45061345 on Build GRCh38
rs752105945 Chr.17: 45061354-45061354 on Build GRCh38
rs759161485 Chr.17: 45061416-45061416 on Build GRCh38
rs946116469 Chr.17: 45061454-45061454 on Build GRCh38
rs764495116 Chr.17: 45061418-45061418 on Build GRCh38
rs575430410 Chr.17: 45061266-45061266 on Build GRCh38
rs575731887 Chr.17: 45061285-45061286 on Build GRCh38
rs973343235 Chr.17: 45061367-45061367 on Build GRCh38
rs770300796 Chr.17: 45061405-45061405 on Build GRCh38
rs776300314 Chr.17: 45061410-45061410 on Build GRCh38
rs1009445042 Chr.17: 45061344-45061344 on Build GRCh38
rs907713546 Chr.17: 45061351-45061351 on Build GRCh38
rs772038652 Chr.17: 45061376-45061376 on Build GRCh38
rs757761311 Chr.17: 45061359-45061359 on Build GRCh38
rs927228181 Chr.17: 45061377-45061377 on Build GRCh38
rs780444510 Chr.17: 45061365-45061365 on Build GRCh38
rs766213031 Chr.17: 45061451-45061453 on Build GRCh38
rs937347334 Chr.17: 45061379-45061379 on Build GRCh38
rs374273277 Chr.17: 45061297-45061297 on Build GRCh38
rs748150811 Chr.17: 45061375-45061375 on Build GRCh38
rs892415557 Chr.17: 45061324-45061324 on Build GRCh38
rs774371000 Chr.17: 45061292-45061292 on Build GRCh38
rs375981533 Chr.17: 45061453-45061455 on Build GRCh38
rs762294220 Chr.17: 45061428-45061428 on Build GRCh38
rs143955814 Chr.17: 45061458-45061458 on Build GRCh38
rs765574685 Chr.17: 45061301-45061301 on Build GRCh38
rs776210749 Chr.17: 45061347-45061347 on Build GRCh38
rs757362692 Chr.17: 45061312-45061312 on Build GRCh38
rs778268422 Chr.17: 45061304-45061304 on Build GRCh38
rs746539074 Chr.17: 45061399-45061399 on Build GRCh38
rs756082410 Chr.17: 45061364-45061364 on Build GRCh38
rs750844557 Chr.17: 45061449-45061449 on Build GRCh38
rs774684452 Chr.17: 45061420-45061420 on Build GRCh38
rs983223652 Chr.17: 45061339-45061339 on Build GRCh38
rs202061902 Chr.17: 45061373-45061373 on Build GRCh38
rs752016137 Chr.17: 45061311-45061311 on Build GRCh38
rs771527303 Chr.17: 45061334-45061334 on Build GRCh38
rs976121906 Chr.17: 45061423-45061423 on Build GRCh38
rs774460908 Chr.17: 45061325-45061325 on Build GRCh38
rs1007666082 Chr.17: 45061362-45061362 on Build GRCh38
rs756247053 Chr.17: 45061451-45061451 on Build GRCh38
rs746125540 Chr.17: 45061313-45061313 on Build GRCh38
rs763334167 Chr.17: 45061350-45061350 on Build GRCh38
rs748067291 Chr.17: 45061327-45061327 on Build GRCh38
rs750501413 Chr.17: 45061286-45061286 on Build GRCh38
rs370358129 Chr.17: 45061293-45061293 on Build GRCh38
rs895881728 Chr.17: 45061427-45061427 on Build GRCh38
rs763044000 Chr.17: 45061283-45061283 on Build GRCh38
rs755233136 Chr.17: 45061466-45061466 on Build GRCh38
rs772952636 Chr.17: 45061388-45061388 on Build GRCh38
rs779782774 Chr.17: 45061316-45061316 on Build GRCh38
rs766535198 Chr.17: 45061455-45061455 on Build GRCh38
rs1018975435 Chr.17: 45061286-45061286 on Build GRCh38
rs754859910 Chr.17: 45061369-45061369 on Build GRCh38
rs750513385 Chr.17: 45061361-45061361 on Build GRCh38
rs369298290 Chr.17: 45061303-45061303 on Build GRCh38
rs767553043 Chr.17: 45061360-45061360 on Build GRCh38
rs768711397 Chr.17: 45061291-45061291 on Build GRCh38
rs763191382 Chr.17: 45061402-45061403 on Build GRCh38
rs772761859 Chr.17: 45061336-45061336 on Build GRCh38

View Full Product Details