Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 68338634-68338859 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
HNRNPH3
Entrez Gene ID
3189
Gene Name
heterogeneous nuclear ribonucleoprotein H3
Gene Aliases
2H9, HNRPH3
UniGene
Hs.643472
Gene Chromosome Location
Chr.10: 68331174-68343196 on Build GRCh38
Transcripts
NM_001322434.1 NM_001322436.1 NM_001322437.1 NM_001322438.1 NM_001322439.1 NM_001322440.1 NM_001322441.1 NM_001322442.1 NM_001322443.1 NM_001322444.1 NM_001322445.1 NM_001322446.1 NM_001322447.1 NM_001322448.1 NM_001322449.1 NM_001322450.1 NM_001322451.1 NM_001322452.1 NM_001322453.1 NM_012207.2 NM_021644.3 XM_011539742.1 XM_017016168.1 XM_017016169.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3415198 c.425G>A p.G142E Chr.10: 68338676-68338676 on Build GRCh38
919680 c.410G>A p.R137Q Chr.10: 68338661-68338661 on Build GRCh38
4015261 c.394A>G p.M132V Chr.10: 68338645-68338645 on Build GRCh38
7195741 c.429_436+5delTGATGGTGGTATG p.? Chr.10: 68338680-68338692 on Build GRCh38
5837171 c.386G>A p.R129H Chr.10: 68338637-68338637 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs970265524 Chr.10: 68338805-68338805 on Build GRCh38
rs199980203 Chr.10: 68338705-68338705 on Build GRCh38
rs987067790 Chr.10: 68338769-68338769 on Build GRCh38
rs867423663 Chr.10: 68338660-68338660 on Build GRCh38
rs779889414 Chr.10: 68338736-68338736 on Build GRCh38
rs200158811 Chr.10: 68338804-68338804 on Build GRCh38
rs959940995 Chr.10: 68338755-68338755 on Build GRCh38
rs1009882618 Chr.10: 68338779-68338791 on Build GRCh38
rs779097052 Chr.10: 68338668-68338668 on Build GRCh38
rs962056808 Chr.10: 68338786-68338786 on Build GRCh38
rs1028640877 Chr.10: 68338732-68338732 on Build GRCh38
rs747875735 Chr.10: 68338710-68338710 on Build GRCh38
rs769595645 Chr.10: 68338714-68338714 on Build GRCh38
rs557506835 Chr.10: 68338742-68338742 on Build GRCh38
rs769812924 Chr.10: 68338729-68338730 on Build GRCh38
rs370270875 Chr.10: 68338706-68338706 on Build GRCh38
rs1031625229 Chr.10: 68338745-68338745 on Build GRCh38
rs772809113 Chr.10: 68338736-68338736 on Build GRCh38
rs538138579 Chr.10: 68338709-68338709 on Build GRCh38
rs746905840 Chr.10: 68338671-68338671 on Build GRCh38
rs774282246 Chr.10: 68338664-68338664 on Build GRCh38
rs111873225 Chr.10: 68338690-68338690 on Build GRCh38
rs993114267 Chr.10: 68338784-68338784 on Build GRCh38
rs972933983 Chr.10: 68338815-68338815 on Build GRCh38
rs754146470 Chr.10: 68338661-68338661 on Build GRCh38
rs757254606 Chr.10: 68338663-68338663 on Build GRCh38

View Full Product Details