Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.X: 64268811-64269070 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
MTMR8
Entrez Gene ID
55613
Gene Name
myotubularin related protein 8
Gene Aliases
-
UniGene
Hs.442892
Gene Chromosome Location
Chr.X: 64268081-64395453 on Build GRCh38
Transcripts
NM_017677.3

SNP Detail

SNP ID Chromosome Location
rs199755703 Chr.X: 64268981-64268981 on Build GRCh38
rs144545484 Chr.X: 64268920-64268920 on Build GRCh38
rs745383604 Chr.X: 64268886-64268886 on Build GRCh38
rs909751044 Chr.X: 64268954-64268954 on Build GRCh38
rs751486345 Chr.X: 64268835-64268835 on Build GRCh38
rs867344788 Chr.X: 64268946-64268946 on Build GRCh38
rs779322916 Chr.X: 64269010-64269010 on Build GRCh38
rs998209132 Chr.X: 64269035-64269035 on Build GRCh38
rs755222027 Chr.X: 64268997-64268997 on Build GRCh38
rs769289024 Chr.X: 64268895-64268895 on Build GRCh38
rs139492339 Chr.X: 64268865-64268865 on Build GRCh38
rs750544576 Chr.X: 64268846-64268846 on Build GRCh38
rs1020188402 Chr.X: 64268890-64268890 on Build GRCh38
rs764053169 Chr.X: 64269018-64269018 on Build GRCh38
rs748960864 Chr.X: 64269046-64269046 on Build GRCh38
rs778280649 Chr.X: 64269019-64269019 on Build GRCh38
rs764840819 Chr.X: 64268877-64268877 on Build GRCh38
rs142319234 Chr.X: 64268836-64268836 on Build GRCh38
rs761785418 Chr.X: 64268925-64268925 on Build GRCh38
rs149703006 Chr.X: 64268868-64268868 on Build GRCh38
rs968678202 Chr.X: 64268893-64268893 on Build GRCh38
rs767568395 Chr.X: 64268931-64268931 on Build GRCh38
rs754057928 Chr.X: 64268980-64268980 on Build GRCh38
rs767695474 Chr.X: 64268838-64268838 on Build GRCh38
rs762557427 Chr.X: 64268909-64268909 on Build GRCh38
rs145287073 Chr.X: 64269011-64269011 on Build GRCh38
rs112395057 Chr.X: 64268991-64268991 on Build GRCh38
rs748650576 Chr.X: 64268869-64268869 on Build GRCh38
rs769319159 Chr.X: 64269031-64269031 on Build GRCh38
rs745330424 Chr.X: 64269030-64269030 on Build GRCh38
rs1003634003 Chr.X: 64268842-64268842 on Build GRCh38
rs760019841 Chr.X: 64268959-64268959 on Build GRCh38
rs139691459 Chr.X: 64268967-64268967 on Build GRCh38
rs771008974 Chr.X: 64269034-64269034 on Build GRCh38
rs766417080 Chr.X: 64268975-64268975 on Build GRCh38
rs546798780 Chr.X: 64269006-64269006 on Build GRCh38
rs150515951 Chr.X: 64268857-64268857 on Build GRCh38
rs760861340 Chr.X: 64268970-64268970 on Build GRCh38
rs147632193 Chr.X: 64269044-64269044 on Build GRCh38
rs768211672 Chr.X: 64269047-64269047 on Build GRCh38
rs756340183 Chr.X: 64268851-64268851 on Build GRCh38
rs752191735 Chr.X: 64268910-64268910 on Build GRCh38
rs780619931 Chr.X: 64268883-64268883 on Build GRCh38
rs775189398 Chr.X: 64268903-64268903 on Build GRCh38

View Full Product Details