Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 97654743-97655000 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
NCBP1
Entrez Gene ID
4686
Gene Name
nuclear cap binding protein subunit 1
Gene Aliases
CBP80, NCBP, Sto1
UniGene
Hs.595669
Gene Chromosome Location
Chr.9: 97633423-97673748 on Build GRCh38
Transcripts
NM_002486.4 XM_011518715.2 XM_017014735.1 XM_017014736.1 XM_017014737.1 XM_017014738.1 XM_006717278.1

Gene Symbol
XPA
Entrez Gene ID
7507
Gene Name
XPA, DNA damage recognition and repair factor
Gene Aliases
XP1, XPAC
UniGene
Hs.654364
Gene Chromosome Location
Chr.9: 97654398-97697409 on Build GRCh38
Transcripts
NM_002486.4 XM_011518715.2 XM_017014735.1 XM_017014736.1 XM_017014737.1 XM_017014738.1 XM_006717278.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6182265 c.1216A>T p.N406Y Chr.9: 97654925-97654925 on Build GRCh38
1103042 c.1228G>A p.V410I Chr.9: 97654937-97654937 on Build GRCh38
1725285 c.1235+1G>T p.? Chr.9: 97654945-97654945 on Build GRCh38
1464329 c.1235+2T>C p.? Chr.9: 97654946-97654946 on Build GRCh38
4609410 c.1174G>T p.A392S Chr.9: 97654883-97654883 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs368988960 Chr.9: 97654956-97654956 on Build GRCh38
rs768012146 Chr.9: 97654860-97654860 on Build GRCh38
rs932772845 Chr.9: 97654813-97654813 on Build GRCh38
rs1048586516 Chr.9: 97654819-97654819 on Build GRCh38
rs774903350 Chr.9: 97654853-97654853 on Build GRCh38
rs1045637221 Chr.9: 97654927-97654927 on Build GRCh38
rs763729923 Chr.9: 97654910-97654910 on Build GRCh38
rs143536712 Chr.9: 97654829-97654829 on Build GRCh38
rs759917197 Chr.9: 97654854-97654854 on Build GRCh38
rs779971908 Chr.9: 97654834-97654834 on Build GRCh38
rs377011060 Chr.9: 97654919-97654919 on Build GRCh38
rs780154642 Chr.9: 97654948-97654948 on Build GRCh38
rs191669699 Chr.9: 97654911-97654911 on Build GRCh38
rs771243403 Chr.9: 97654846-97654846 on Build GRCh38
rs758345054 Chr.9: 97654942-97654942 on Build GRCh38
rs895448217 Chr.9: 97654961-97654961 on Build GRCh38
rs1056823550 Chr.9: 97654957-97654957 on Build GRCh38
rs866467789 Chr.9: 97654802-97654802 on Build GRCh38
rs757052791 Chr.9: 97654916-97654916 on Build GRCh38
rs750263253 Chr.9: 97654933-97654933 on Build GRCh38
rs746981723 Chr.9: 97654903-97654903 on Build GRCh38
rs561870475 Chr.9: 97654823-97654823 on Build GRCh38
rs749662440 Chr.9: 97654840-97654840 on Build GRCh38
rs1004451067 Chr.9: 97654827-97654827 on Build GRCh38
rs748226866 Chr.9: 97654955-97654955 on Build GRCh38
rs900003885 Chr.9: 97654778-97654778 on Build GRCh38
rs906507720 Chr.9: 97654872-97654872 on Build GRCh38
rs201715578 Chr.9: 97654907-97654907 on Build GRCh38

View Full Product Details