Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 53930014-53930261 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
HSPB11
Entrez Gene ID
51668
Gene Name
heat shock protein family B (small) member 11
Gene Aliases
C1orf41, HSPCO34, IFT25, PP25
UniGene
Hs.525462
Gene Chromosome Location
Chr.1: 53921561-53946308 on Build GRCh38
Transcripts
NM_001316935.1 NM_016126.3 NR_133632.1 NR_133633.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6385033 c.195A>T p.Q65H Chr.1: 53930049-53930049 on Build GRCh38
6385032 c.195A>T p.Q65H Chr.1: 53930049-53930049 on Build GRCh38
414844 c.110C>T p.T37M Chr.1: 53930134-53930134 on Build GRCh38
4008908 c.131T>C p.M44T Chr.1: 53930113-53930113 on Build GRCh38
3985142 c.160C>T p.H54Y Chr.1: 53930084-53930084 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs188148398 Chr.1: 53930232-53930232 on Build GRCh38
rs763883555 Chr.1: 53930186-53930186 on Build GRCh38
rs780679621 Chr.1: 53930123-53930123 on Build GRCh38
rs774579110 Chr.1: 53930141-53930141 on Build GRCh38
rs369947892 Chr.1: 53930133-53930133 on Build GRCh38
rs775392074 Chr.1: 53930181-53930181 on Build GRCh38
rs183116954 Chr.1: 53930217-53930217 on Build GRCh38
rs191768908 Chr.1: 53930215-53930215 on Build GRCh38
rs759014261 Chr.1: 53930175-53930175 on Build GRCh38
rs751343618 Chr.1: 53930190-53930190 on Build GRCh38
rs777486814 Chr.1: 53930077-53930077 on Build GRCh38
rs747028464 Chr.1: 53930196-53930196 on Build GRCh38
rs756313870 Chr.1: 53930097-53930097 on Build GRCh38
rs745958906 Chr.1: 53930096-53930096 on Build GRCh38
rs186020159 Chr.1: 53930169-53930169 on Build GRCh38
rs759417292 Chr.1: 53930050-53930050 on Build GRCh38
rs781293591 Chr.1: 53930083-53930083 on Build GRCh38
rs1001857482 Chr.1: 53930236-53930236 on Build GRCh38
rs970025858 Chr.1: 53930235-53930235 on Build GRCh38
rs377492199 Chr.1: 53930176-53930176 on Build GRCh38
rs762891668 Chr.1: 53930183-53930183 on Build GRCh38
rs200563992 Chr.1: 53930061-53930061 on Build GRCh38
rs752670381 Chr.1: 53930057-53930057 on Build GRCh38
rs531248700 Chr.1: 53930052-53930052 on Build GRCh38
rs769134105 Chr.1: 53930134-53930134 on Build GRCh38
rs536718437 Chr.1: 53930145-53930145 on Build GRCh38
rs746506620 Chr.1: 53930061-53930062 on Build GRCh38
rs758184922 Chr.1: 53930076-53930076 on Build GRCh38
rs776391708 Chr.1: 53930173-53930173 on Build GRCh38
rs757425229 Chr.1: 53930079-53930079 on Build GRCh38
rs746628902 Chr.1: 53930149-53930149 on Build GRCh38
rs547575301 Chr.1: 53930218-53930218 on Build GRCh38

View Full Product Details