Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 45924753-45924980 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
LARGE2
Entrez Gene ID
120071
Gene Name
LARGE xylosyl- and glucuronyltransferase 2
Gene Aliases
GYLTL1B, PP5656
UniGene
Hs.86543
Gene Chromosome Location
Chr.11: 45921621-45929096 on Build GRCh38
Transcripts
NM_001300721.1 NM_001300722.1 NM_152312.4 XM_005252787.2 XM_006718140.1 XM_006718141.3 XM_011519886.1 XM_011519888.1 XM_011519889.1 XM_011519890.1 XM_011519891.1 XM_011519892.2 XM_011519893.1 XM_017017173.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4977164 c.702C>T p.D234D Chr.11: 45924822-45924822 on Build GRCh38
1353895 c.701A>G p.D234G Chr.11: 45924821-45924821 on Build GRCh38
5372494 c.712G>A p.G238S Chr.11: 45924832-45924832 on Build GRCh38
927098 c.674C>T p.A225V Chr.11: 45924794-45924794 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs931869979 Chr.11: 45924828-45924828 on Build GRCh38
rs925745022 Chr.11: 45924922-45924922 on Build GRCh38
rs923088326 Chr.11: 45924801-45924801 on Build GRCh38
rs775582149 Chr.11: 45924788-45924788 on Build GRCh38
rs776687854 Chr.11: 45924849-45924849 on Build GRCh38
rs12576236 Chr.11: 45924821-45924821 on Build GRCh38
rs552600788 Chr.11: 45924881-45924881 on Build GRCh38
rs763585348 Chr.11: 45924894-45924894 on Build GRCh38
rs747695475 Chr.11: 45924795-45924795 on Build GRCh38
rs779334648 Chr.11: 45924921-45924921 on Build GRCh38
rs866210861 Chr.11: 45924807-45924807 on Build GRCh38
rs750731745 Chr.11: 45924892-45924892 on Build GRCh38
rs975853295 Chr.11: 45924800-45924800 on Build GRCh38
rs941993848 Chr.11: 45924876-45924876 on Build GRCh38
rs548421543 Chr.11: 45924941-45924941 on Build GRCh38
rs368096853 Chr.11: 45924918-45924918 on Build GRCh38
rs972951261 Chr.11: 45924774-45924774 on Build GRCh38
rs57937599 Chr.11: 45924882-45924882 on Build GRCh38
rs189478803 Chr.11: 45924919-45924919 on Build GRCh38
rs189140907 Chr.11: 45924917-45924917 on Build GRCh38
rs532814183 Chr.11: 45924843-45924843 on Build GRCh38
rs942761616 Chr.11: 45924905-45924905 on Build GRCh38
rs909135231 Chr.11: 45924875-45924875 on Build GRCh38
rs762307296 Chr.11: 45924852-45924852 on Build GRCh38
rs146900807 Chr.11: 45924855-45924855 on Build GRCh38
rs760698896 Chr.11: 45924796-45924796 on Build GRCh38

View Full Product Details