Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 69915348-69915597 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
MXD1
Entrez Gene ID
4084
Gene Name
MAX dimerization protein 1
Gene Aliases
BHLHC58, MAD, MAD1
UniGene
Hs.468908
Gene Chromosome Location
Chr.2: 69915041-69942945 on Build GRCh38
Transcripts
NM_001202513.1 NM_001202514.1 NM_002357.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1327001 c.42G>A p.E14E Chr.2: 69915372-69915372 on Build GRCh38
1185928 c.68A>G p.E23G Chr.2: 69915398-69915398 on Build GRCh38
6431827 c.38T>C p.L13P Chr.2: 69915368-69915368 on Build GRCh38
1409159 c.60G>A p.E20E Chr.2: 69915390-69915390 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs541449740 Chr.2: 69915463-69915463 on Build GRCh38
rs892217199 Chr.2: 69915419-69915419 on Build GRCh38
rs1009195901 Chr.2: 69915447-69915447 on Build GRCh38
rs1033471786 Chr.2: 69915521-69915521 on Build GRCh38
rs866428049 Chr.2: 69915446-69915446 on Build GRCh38
rs927437467 Chr.2: 69915475-69915475 on Build GRCh38
rs979869432 Chr.2: 69915468-69915468 on Build GRCh38
rs1030282836 Chr.2: 69915394-69915394 on Build GRCh38
rs750350464 Chr.2: 69915418-69915418 on Build GRCh38
rs386647105 Chr.2: 69915407-69915408 on Build GRCh38
rs779736964 Chr.2: 69915449-69915449 on Build GRCh38
rs555686751 Chr.2: 69915420-69915420 on Build GRCh38
rs903739574 Chr.2: 69915451-69915451 on Build GRCh38
rs757721297 Chr.2: 69915502-69915502 on Build GRCh38
rs950361116 Chr.2: 69915429-69915429 on Build GRCh38
rs891330513 Chr.2: 69915407-69915407 on Build GRCh38
rs564302289 Chr.2: 69915556-69915556 on Build GRCh38
rs1051280353 Chr.2: 69915405-69915405 on Build GRCh38
rs1002100761 Chr.2: 69915505-69915505 on Build GRCh38
rs572639550 Chr.2: 69915423-69915423 on Build GRCh38
rs1057502246 Chr.2: 69915492-69915492 on Build GRCh38
rs1039813572 Chr.2: 69915576-69915576 on Build GRCh38
rs991962678 Chr.2: 69915537-69915537 on Build GRCh38
rs962459930 Chr.2: 69915571-69915571 on Build GRCh38
rs938889015 Chr.2: 69915490-69915490 on Build GRCh38
rs960563215 Chr.2: 69915529-69915529 on Build GRCh38
rs1039829401 Chr.2: 69915425-69915425 on Build GRCh38
rs1006905672 Chr.2: 69915574-69915574 on Build GRCh38
rs879613430 Chr.2: 69915566-69915566 on Build GRCh38
rs71413200 Chr.2: 69915500-69915500 on Build GRCh38
rs930088133 Chr.2: 69915398-69915398 on Build GRCh38
rs941733309 Chr.2: 69915466-69915466 on Build GRCh38

View Full Product Details