Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 130987178-130987445 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ADGRD1
Entrez Gene ID
283383
Gene Name
adhesion G protein-coupled receptor D1
Gene Aliases
GPR133, PGR25
UniGene
Hs.719239
Gene Chromosome Location
Chr.12: 130953907-131141469 on Build GRCh38
Transcripts
NM_198827.3 XM_005253566.2 XM_011538203.2 XM_011538204.2 XM_011538205.2 XM_011538206.2 XM_011538207.2 XM_011538208.2 XM_011538209.2 XM_011538210.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7280242 c.706A>T p.T236S Chr.12: 130987310-130987310 on Build GRCh38
6834943 c.698G>A p.R233Q Chr.12: 130987302-130987302 on Build GRCh38
546982 c.578C>T p.P193L Chr.12: 130987182-130987182 on Build GRCh38
3792361 c.679G>A p.E227K Chr.12: 130987283-130987283 on Build GRCh38
2228555 c.675C>T p.F225F Chr.12: 130987279-130987279 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs199837477 Chr.12: 130987315-130987315 on Build GRCh38
rs897376702 Chr.12: 130987403-130987403 on Build GRCh38
rs759367752 Chr.12: 130987271-130987271 on Build GRCh38
rs764239492 Chr.12: 130987282-130987282 on Build GRCh38
rs745958911 Chr.12: 130987219-130987219 on Build GRCh38
rs749549808 Chr.12: 130987314-130987314 on Build GRCh38
rs2398544 Chr.12: 130987411-130987411 on Build GRCh38
rs780500944 Chr.12: 130987309-130987309 on Build GRCh38
rs56674647 Chr.12: 130987412-130987412 on Build GRCh38
rs146191332 Chr.12: 130987388-130987388 on Build GRCh38
rs772855107 Chr.12: 130987259-130987259 on Build GRCh38
rs746461245 Chr.12: 130987264-130987264 on Build GRCh38
rs762710525 Chr.12: 130987370-130987370 on Build GRCh38
rs763022510 Chr.12: 130987315-130987315 on Build GRCh38
rs951078264 Chr.12: 130987235-130987235 on Build GRCh38
rs76898752 Chr.12: 130987414-130987414 on Build GRCh38
rs201057341 Chr.12: 130987329-130987329 on Build GRCh38
rs751764117 Chr.12: 130987284-130987284 on Build GRCh38
rs139478688 Chr.12: 130987229-130987229 on Build GRCh38
rs750433039 Chr.12: 130987391-130987391 on Build GRCh38
rs763867099 Chr.12: 130987371-130987371 on Build GRCh38
rs368468703 Chr.12: 130987325-130987325 on Build GRCh38
rs377562590 Chr.12: 130987280-130987280 on Build GRCh38
rs765101760 Chr.12: 130987272-130987272 on Build GRCh38
rs770561309 Chr.12: 130987266-130987266 on Build GRCh38
rs756531633 Chr.12: 130987305-130987305 on Build GRCh38
rs202010208 Chr.12: 130987279-130987279 on Build GRCh38
rs975856156 Chr.12: 130987239-130987239 on Build GRCh38
rs781710819 Chr.12: 130987217-130987217 on Build GRCh38
rs776749565 Chr.12: 130987270-130987270 on Build GRCh38
rs541028270 Chr.12: 130987356-130987356 on Build GRCh38
rs774600391 Chr.12: 130987378-130987378 on Build GRCh38
rs766740494 Chr.12: 130987397-130987397 on Build GRCh38
rs201053759 Chr.12: 130987338-130987338 on Build GRCh38
rs201045213 Chr.12: 130987213-130987213 on Build GRCh38
rs775365854 Chr.12: 130987358-130987358 on Build GRCh38
rs764268842 Chr.12: 130987208-130987208 on Build GRCh38
rs375084180 Chr.12: 130987200-130987200 on Build GRCh38
rs770612786 Chr.12: 130987327-130987327 on Build GRCh38
rs370807516 Chr.12: 130987301-130987301 on Build GRCh38
rs747860414 Chr.12: 130987225-130987225 on Build GRCh38
rs766544060 Chr.12: 130987369-130987370 on Build GRCh38
rs746373801 Chr.12: 130987218-130987218 on Build GRCh38
rs562234701 Chr.12: 130987324-130987324 on Build GRCh38
rs757399520 Chr.12: 130987291-130987291 on Build GRCh38
rs761251069 Chr.12: 130987395-130987395 on Build GRCh38
rs750980141 Chr.12: 130987302-130987302 on Build GRCh38
rs945360352 Chr.12: 130987419-130987419 on Build GRCh38
rs758503212 Chr.12: 130987201-130987201 on Build GRCh38
rs749755466 Chr.12: 130987365-130987365 on Build GRCh38
rs372724799 Chr.12: 130987380-130987380 on Build GRCh38

View Full Product Details