Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 46039493-46039750 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
KANSL1
Entrez Gene ID
284058
Gene Name
KAT8 regulatory NSL complex subunit 1
Gene Aliases
CENP-36, KDVS, KIAA1267, MSL1v1, NSL1, hMSL1v1
UniGene
Hs.648744
Gene Chromosome Location
Chr.17: 46029916-46225374 on Build GRCh38
Transcripts
NM_001193465.1 NM_001193466.1 NM_015443.3 XM_006721823.1 XM_006721824.3 XM_011524628.2 XM_011524629.2 XM_011524630.2 XM_011524631.2 XM_011524632.2 XM_017024488.1 XM_017024489.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5566736 c.2160T>G p.R720R Chr.17: 46039745-46039745 on Build GRCh38
436742 c.2158delC p.R720fs*12 Chr.17: 46039747-46039747 on Build GRCh38
336074 c.2181C>T p.V727V Chr.17: 46039724-46039724 on Build GRCh38
7002453 c.2159_2160insCG p.K721fs*12 Chr.17: 46039745-46039746 on Build GRCh38
5904637 c.2158C>T p.R720C Chr.17: 46039747-46039747 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs561916424 Chr.17: 46039609-46039609 on Build GRCh38
rs919683693 Chr.17: 46039578-46039578 on Build GRCh38
rs1015202385 Chr.17: 46039548-46039548 on Build GRCh38
rs940435605 Chr.17: 46039680-46039680 on Build GRCh38
rs544672172 Chr.17: 46039590-46039590 on Build GRCh38
rs796052595 Chr.17: 46039701-46039701 on Build GRCh38
rs572992096 Chr.17: 46039588-46039588 on Build GRCh38
rs542054201 Chr.17: 46039589-46039589 on Build GRCh38
rs150150664 Chr.17: 46039627-46039627 on Build GRCh38
rs762283385 Chr.17: 46039690-46039690 on Build GRCh38
rs958563180 Chr.17: 46039576-46039576 on Build GRCh38
rs544583287 Chr.17: 46039637-46039637 on Build GRCh38
rs893210793 Chr.17: 46039631-46039631 on Build GRCh38
rs766092464 Chr.17: 46039654-46039654 on Build GRCh38
rs1042351150 Chr.17: 46039603-46039604 on Build GRCh38
rs145434865 Chr.17: 46039715-46039715 on Build GRCh38
rs973827735 Chr.17: 46039569-46039569 on Build GRCh38
rs1028441222 Chr.17: 46039653-46039653 on Build GRCh38
rs995181923 Chr.17: 46039543-46039543 on Build GRCh38
rs934670157 Chr.17: 46039621-46039621 on Build GRCh38
rs934186127 Chr.17: 46039647-46039647 on Build GRCh38
rs764721407 Chr.17: 46039684-46039684 on Build GRCh38
rs949268716 Chr.17: 46039595-46039595 on Build GRCh38
rs374338680 Chr.17: 46039675-46039675 on Build GRCh38
rs369133174 Chr.17: 46039663-46039663 on Build GRCh38
rs957373107 Chr.17: 46039620-46039620 on Build GRCh38
rs1034254371 Chr.17: 46039559-46039559 on Build GRCh38
rs780291785 Chr.17: 46039727-46039727 on Build GRCh38
rs564357484 Chr.17: 46039649-46039649 on Build GRCh38
rs867261416 Chr.17: 46039527-46039527 on Build GRCh38
rs925963213 Chr.17: 46039610-46039610 on Build GRCh38
rs80026189 Chr.17: 46039708-46039708 on Build GRCh38
rs898802995 Chr.17: 46039533-46039533 on Build GRCh38
rs1036089094 Chr.17: 46039711-46039711 on Build GRCh38
rs7221390 Chr.17: 46039584-46039584 on Build GRCh38
rs372797286 Chr.17: 46039685-46039685 on Build GRCh38
rs1015013203 Chr.17: 46039528-46039528 on Build GRCh38
rs987049793 Chr.17: 46039547-46039547 on Build GRCh38

View Full Product Details