Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 43386859-43387106 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
HNRNPF
Entrez Gene ID
3185
Gene Name
heterogeneous nuclear ribonucleoprotein F
Gene Aliases
HNRPF, OK/SW-cl.23, mcs94-1
UniGene
Hs.712955
Gene Chromosome Location
Chr.10: 43385617-43409248 on Build GRCh38
Transcripts
NM_001098204.1 NM_001098205.1 NM_001098206.1 NM_001098207.1 NM_001098208.1 NM_004966.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
918166 c.990A>G p.E330E Chr.10: 43386895-43386895 on Build GRCh38
4014197 c.895A>G p.K299E Chr.10: 43386990-43386990 on Build GRCh38
918167 c.990A>G p.E330E Chr.10: 43386895-43386895 on Build GRCh38
4014196 c.895A>G p.K299E Chr.10: 43386990-43386990 on Build GRCh38
4014194 c.1009A>G p.T337A Chr.10: 43386876-43386876 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs376527546 Chr.10: 43387063-43387063 on Build GRCh38
rs17849827 Chr.10: 43387075-43387075 on Build GRCh38
rs768196856 Chr.10: 43387031-43387031 on Build GRCh38
rs963637550 Chr.10: 43386923-43386923 on Build GRCh38
rs749516664 Chr.10: 43387074-43387074 on Build GRCh38
rs759602508 Chr.10: 43387077-43387077 on Build GRCh38
rs146750316 Chr.10: 43387081-43387081 on Build GRCh38
rs746441995 Chr.10: 43387018-43387018 on Build GRCh38
rs566452971 Chr.10: 43387054-43387054 on Build GRCh38
rs768407887 Chr.10: 43386898-43386898 on Build GRCh38
rs151043426 Chr.10: 43386982-43386982 on Build GRCh38
rs879548216 Chr.10: 43386949-43386949 on Build GRCh38
rs142085844 Chr.10: 43386985-43386985 on Build GRCh38
rs756589564 Chr.10: 43386986-43386986 on Build GRCh38
rs763266513 Chr.10: 43387060-43387060 on Build GRCh38
rs993397756 Chr.10: 43386976-43386976 on Build GRCh38
rs533557195 Chr.10: 43387056-43387056 on Build GRCh38
rs554278003 Chr.10: 43386994-43386994 on Build GRCh38
rs772941210 Chr.10: 43387057-43387057 on Build GRCh38
rs756743272 Chr.10: 43386946-43386946 on Build GRCh38
rs779624717 Chr.10: 43387010-43387010 on Build GRCh38
rs755522714 Chr.10: 43386975-43386975 on Build GRCh38
rs201947175 Chr.10: 43387072-43387072 on Build GRCh38
rs766502001 Chr.10: 43386930-43386930 on Build GRCh38
rs750681992 Chr.10: 43386924-43386924 on Build GRCh38
rs149892317 Chr.10: 43386925-43386925 on Build GRCh38
rs765009128 Chr.10: 43386922-43386922 on Build GRCh38
rs761651144 Chr.10: 43386902-43386902 on Build GRCh38
rs776613768 Chr.10: 43387032-43387032 on Build GRCh38
rs771588483 Chr.10: 43387001-43387001 on Build GRCh38
rs140104128 Chr.10: 43386901-43386901 on Build GRCh38
rs755210151 Chr.10: 43387078-43387079 on Build GRCh38
rs559011933 Chr.10: 43386970-43386970 on Build GRCh38
rs144952315 Chr.10: 43386961-43386961 on Build GRCh38
rs756642926 Chr.10: 43387083-43387083 on Build GRCh38
rs767675065 Chr.10: 43387078-43387078 on Build GRCh38

View Full Product Details