Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 44249344-44249617 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
HSP90AB1
Entrez Gene ID
3326
Gene Name
heat shock protein 90 alpha family class B member 1
Gene Aliases
D6S182, HSP84, HSP90B, HSPC2, HSPCB
UniGene
Hs.509736
Gene Chromosome Location
Chr.6: 44246166-44253888 on Build GRCh38
Transcripts
NM_001271969.1 NM_001271970.1 NM_001271971.1 NM_001271972.1 NM_007355.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6682495 c.245G>A p.R82H Chr.6: 44249474-44249474 on Build GRCh38
742825 c.177G>C p.L59L Chr.6: 44249406-44249406 on Build GRCh38
6336471 c.189G>A p.S63S Chr.6: 44249418-44249418 on Build GRCh38
7110276 c.258G>A p.L86L Chr.6: 44249487-44249487 on Build GRCh38
320883 c.301A>T p.N101Y Chr.6: 44249530-44249530 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs61753592 Chr.6: 44249473-44249473 on Build GRCh38
rs138497879 Chr.6: 44249423-44249423 on Build GRCh38
rs778973317 Chr.6: 44249418-44249418 on Build GRCh38
rs775163914 Chr.6: 44249466-44249466 on Build GRCh38
rs147289632 Chr.6: 44249521-44249521 on Build GRCh38
rs143977102 Chr.6: 44249463-44249463 on Build GRCh38
rs11538987 Chr.6: 44249479-44249479 on Build GRCh38
rs766692041 Chr.6: 44249374-44249374 on Build GRCh38
rs761163017 Chr.6: 44249485-44249485 on Build GRCh38
rs777662526 Chr.6: 44249430-44249430 on Build GRCh38
rs149441958 Chr.6: 44249397-44249397 on Build GRCh38
rs1039547052 Chr.6: 44249462-44249462 on Build GRCh38
rs771117200 Chr.6: 44249396-44249396 on Build GRCh38
rs918222067 Chr.6: 44249422-44249422 on Build GRCh38
rs776401249 Chr.6: 44249450-44249450 on Build GRCh38
rs777752128 Chr.6: 44249547-44249547 on Build GRCh38
rs758350543 Chr.6: 44249427-44249427 on Build GRCh38
rs372668329 Chr.6: 44249488-44249488 on Build GRCh38
rs755172327 Chr.6: 44249417-44249417 on Build GRCh38
rs201247226 Chr.6: 44249540-44249540 on Build GRCh38
rs954810817 Chr.6: 44249459-44249459 on Build GRCh38
rs751437633 Chr.6: 44249574-44249574 on Build GRCh38
rs552784311 Chr.6: 44249514-44249514 on Build GRCh38
rs770625988 Chr.6: 44249442-44249442 on Build GRCh38
rs909623121 Chr.6: 44249452-44249452 on Build GRCh38
rs765391115 Chr.6: 44249524-44249524 on Build GRCh38
rs373969944 Chr.6: 44249469-44249469 on Build GRCh38
rs766669218 Chr.6: 44249511-44249511 on Build GRCh38
rs746857821 Chr.6: 44249438-44249438 on Build GRCh38
rs11538982 Chr.6: 44249460-44249460 on Build GRCh38
rs987570357 Chr.6: 44249464-44249464 on Build GRCh38
rs201634463 Chr.6: 44249472-44249472 on Build GRCh38
rs752819438 Chr.6: 44249531-44249531 on Build GRCh38
rs11538983 Chr.6: 44249394-44249394 on Build GRCh38
rs747643899 Chr.6: 44249456-44249456 on Build GRCh38
rs1052451895 Chr.6: 44249476-44249476 on Build GRCh38
rs147535516 Chr.6: 44249413-44249413 on Build GRCh38

View Full Product Details