Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 36954618-36954889 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PI16
Entrez Gene ID
221476
Gene Name
peptidase inhibitor 16
Gene Aliases
CD364, CRISP9, MSMBBP, PSPBP
UniGene
Hs.25391
Gene Chromosome Location
Chr.6: 36948263-36964837 on Build GRCh38
Transcripts
NM_001199159.1 NM_153370.2 XM_005248917.2 XM_011514375.2 XM_017010430.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5405152 c.33G>A p.L11L Chr.6: 36954793-36954793 on Build GRCh38
742167 c.91G>T p.E31* Chr.6: 36954851-36954851 on Build GRCh38
5912483 c.94G>A p.E32K Chr.6: 36954854-36954854 on Build GRCh38
742168 c.70C>A p.P24T Chr.6: 36954830-36954830 on Build GRCh38
4557762 c.73G>A p.V25I Chr.6: 36954833-36954833 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs766298919 Chr.6: 36954799-36954799 on Build GRCh38
rs779382265 Chr.6: 36954825-36954825 on Build GRCh38
rs754397097 Chr.6: 36954816-36954816 on Build GRCh38
rs1018610606 Chr.6: 36954850-36954850 on Build GRCh38
rs202171420 Chr.6: 36954766-36954766 on Build GRCh38
rs774354838 Chr.6: 36954669-36954669 on Build GRCh38
rs116584983 Chr.6: 36954778-36954778 on Build GRCh38
rs925039823 Chr.6: 36954662-36954662 on Build GRCh38
rs758589274 Chr.6: 36954785-36954785 on Build GRCh38
rs780601185 Chr.6: 36954727-36954727 on Build GRCh38
rs1534176 Chr.6: 36954747-36954747 on Build GRCh38
rs759353266 Chr.6: 36954752-36954752 on Build GRCh38
rs898762504 Chr.6: 36954639-36954639 on Build GRCh38
rs148879002 Chr.6: 36954832-36954832 on Build GRCh38
rs200171043 Chr.6: 36954734-36954734 on Build GRCh38
rs762064253 Chr.6: 36954773-36954773 on Build GRCh38
rs772887864 Chr.6: 36954744-36954744 on Build GRCh38
rs756685181 Chr.6: 36954725-36954725 on Build GRCh38
rs755388075 Chr.6: 36954819-36954819 on Build GRCh38
rs770226171 Chr.6: 36954738-36954740 on Build GRCh38
rs146508132 Chr.6: 36954863-36954863 on Build GRCh38
rs749696863 Chr.6: 36954730-36954730 on Build GRCh38
rs932501296 Chr.6: 36954798-36954798 on Build GRCh38
rs760235156 Chr.6: 36954749-36954749 on Build GRCh38
rs144573705 Chr.6: 36954845-36954845 on Build GRCh38
rs750564019 Chr.6: 36954777-36954777 on Build GRCh38
rs746593536 Chr.6: 36954834-36954834 on Build GRCh38
rs144930102 Chr.6: 36954767-36954767 on Build GRCh38
rs761512620 Chr.6: 36954798-36954799 on Build GRCh38
rs770290491 Chr.6: 36954750-36954750 on Build GRCh38
rs1033813689 Chr.6: 36954675-36954675 on Build GRCh38
rs375168502 Chr.6: 36954751-36954751 on Build GRCh38
rs143532329 Chr.6: 36954833-36954833 on Build GRCh38
rs775334493 Chr.6: 36954860-36954860 on Build GRCh38
rs752250405 Chr.6: 36954737-36954737 on Build GRCh38
rs987017731 Chr.6: 36954801-36954801 on Build GRCh38
rs767725611 Chr.6: 36954776-36954776 on Build GRCh38
rs750694256 Chr.6: 36954735-36954735 on Build GRCh38
rs971536676 Chr.6: 36954742-36954742 on Build GRCh38
rs1014165393 Chr.6: 36954691-36954691 on Build GRCh38
rs995683535 Chr.6: 36954643-36954643 on Build GRCh38
rs753079501 Chr.6: 36954826-36954826 on Build GRCh38
rs185446579 Chr.6: 36954689-36954689 on Build GRCh38
rs201111148 Chr.6: 36954732-36954732 on Build GRCh38
rs148119018 Chr.6: 36954841-36954841 on Build GRCh38
rs769590040 Chr.6: 36954852-36954852 on Build GRCh38

View Full Product Details