Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 51716923-51717180 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
HAS1
Entrez Gene ID
3036
Gene Name
hyaluronan synthase 1
Gene Aliases
HAS
UniGene
Hs.57697
Gene Chromosome Location
Chr.19: 51713112-51723992 on Build GRCh38
Transcripts
NM_001297436.1 NM_001523.3 XM_011526884.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7223156 c.874G>C p.E292Q Chr.19: 51717022-51717022 on Build GRCh38
5651187 c.880G>A p.A294T Chr.19: 51717016-51717016 on Build GRCh38
372956 c.775C>A p.R259R Chr.19: 51717121-51717121 on Build GRCh38
713101 c.851G>C p.R284P Chr.19: 51717045-51717045 on Build GRCh38
3537875 c.785C>T p.A262V Chr.19: 51717111-51717111 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs776266210 Chr.19: 51717044-51717044 on Build GRCh38
rs541103037 Chr.19: 51716986-51716986 on Build GRCh38
rs140416890 Chr.19: 51716972-51716972 on Build GRCh38
rs758033599 Chr.19: 51716957-51716957 on Build GRCh38
rs372673212 Chr.19: 51717118-51717118 on Build GRCh38
rs1043616888 Chr.19: 51717007-51717007 on Build GRCh38
rs866172072 Chr.19: 51717076-51717076 on Build GRCh38
rs749715354 Chr.19: 51716977-51716977 on Build GRCh38
rs764755611 Chr.19: 51717109-51717109 on Build GRCh38
rs747392663 Chr.19: 51717131-51717131 on Build GRCh38
rs1007149481 Chr.19: 51716994-51716994 on Build GRCh38
rs780774711 Chr.19: 51716976-51716976 on Build GRCh38
rs1030207517 Chr.19: 51716950-51716950 on Build GRCh38
rs759007645 Chr.19: 51717130-51717130 on Build GRCh38
rs75142551 Chr.19: 51717141-51717141 on Build GRCh38
rs773180567 Chr.19: 51717113-51717113 on Build GRCh38
rs948729841 Chr.19: 51717094-51717094 on Build GRCh38
rs201445042 Chr.19: 51717146-51717146 on Build GRCh38
rs754654499 Chr.19: 51717079-51717079 on Build GRCh38
rs11084111 Chr.19: 51717098-51717098 on Build GRCh38
rs779268942 Chr.19: 51717123-51717123 on Build GRCh38
rs200078680 Chr.19: 51717045-51717045 on Build GRCh38
rs756579719 Chr.19: 51717010-51717010 on Build GRCh38
rs766119804 Chr.19: 51717119-51717119 on Build GRCh38
rs746297042 Chr.19: 51716997-51716997 on Build GRCh38
rs768751929 Chr.19: 51717037-51717037 on Build GRCh38
rs750914461 Chr.19: 51717125-51717125 on Build GRCh38
rs757687431 Chr.19: 51717086-51717086 on Build GRCh38
rs751162741 Chr.19: 51717067-51717067 on Build GRCh38
rs768928691 Chr.19: 51717097-51717097 on Build GRCh38
rs765234716 Chr.19: 51717027-51717027 on Build GRCh38
rs765992957 Chr.19: 51717047-51717047 on Build GRCh38
rs1052249811 Chr.19: 51717026-51717026 on Build GRCh38
rs752261227 Chr.19: 51717083-51717083 on Build GRCh38
rs781516429 Chr.19: 51717142-51717142 on Build GRCh38
rs1032247937 Chr.19: 51717033-51717033 on Build GRCh38
rs150290681 Chr.19: 51717018-51717018 on Build GRCh38
rs956193883 Chr.19: 51717054-51717054 on Build GRCh38
rs779544729 Chr.19: 51717089-51717089 on Build GRCh38
rs780498466 Chr.19: 51717081-51717081 on Build GRCh38
rs760818172 Chr.19: 51717005-51717005 on Build GRCh38
rs910008923 Chr.19: 51717102-51717102 on Build GRCh38
rs772045482 Chr.19: 51717117-51717118 on Build GRCh38
rs772422528 Chr.19: 51717002-51717002 on Build GRCh38
rs746490264 Chr.19: 51716960-51716960 on Build GRCh38
rs201369230 Chr.19: 51717004-51717004 on Build GRCh38
rs147158919 Chr.19: 51717121-51717121 on Build GRCh38
rs137939869 Chr.19: 51717093-51717093 on Build GRCh38
rs750095254 Chr.19: 51717145-51717145 on Build GRCh38
rs771470487 Chr.19: 51716979-51716979 on Build GRCh38
rs779748601 Chr.19: 51716958-51716958 on Build GRCh38
rs554817156 Chr.19: 51717112-51717112 on Build GRCh38
rs770866299 Chr.19: 51717160-51717160 on Build GRCh38
rs78761398 Chr.19: 51717148-51717148 on Build GRCh38
rs530794115 Chr.19: 51717120-51717120 on Build GRCh38
rs772616275 Chr.19: 51717091-51717091 on Build GRCh38
rs762776728 Chr.19: 51717046-51717046 on Build GRCh38
rs370359788 Chr.19: 51716942-51716942 on Build GRCh38
rs761590658 Chr.19: 51717019-51717019 on Build GRCh38
rs750236662 Chr.19: 51716954-51716954 on Build GRCh38
rs755693950 Chr.19: 51717122-51717122 on Build GRCh38
rs1052127701 Chr.19: 51717126-51717126 on Build GRCh38
rs746207145 Chr.19: 51717090-51717090 on Build GRCh38

View Full Product Details