Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 945515-945768 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
NOC2L
Entrez Gene ID
26155
Gene Name
NOC2 like nucleolar associated transcriptional repressor
Gene Aliases
NET15, NET7, NIR, PPP1R112
UniGene
Hs.405987
Gene Chromosome Location
Chr.1: 944203-959299 on Build GRCh38
Transcripts
NM_015658.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1296758 c.2026G>A p.D676N Chr.1: 945545-945545 on Build GRCh38
4707442 c.1983G>A p.R661R Chr.1: 945588-945588 on Build GRCh38
5535508 c.2033C>T p.T678I Chr.1: 945538-945538 on Build GRCh38
5790652 c.2001C>G p.L667L Chr.1: 945570-945570 on Build GRCh38
5686930 c.1929G>T p.L643L Chr.1: 945642-945642 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs951219625 Chr.1: 945620-945623 on Build GRCh38
rs776848413 Chr.1: 945673-945673 on Build GRCh38
rs369553946 Chr.1: 945623-945623 on Build GRCh38
rs1045144424 Chr.1: 945721-945721 on Build GRCh38
rs767240804 Chr.1: 945625-945625 on Build GRCh38
rs779361120 Chr.1: 945568-945568 on Build GRCh38
rs772663096 Chr.1: 945536-945536 on Build GRCh38
rs200256358 Chr.1: 945675-945675 on Build GRCh38
rs745985410 Chr.1: 945657-945658 on Build GRCh38
rs779464694 Chr.1: 945585-945585 on Build GRCh38
rs751334404 Chr.1: 945550-945550 on Build GRCh38
rs368403481 Chr.1: 945630-945630 on Build GRCh38
rs377368061 Chr.1: 945660-945660 on Build GRCh38
rs753261930 Chr.1: 945658-945658 on Build GRCh38
rs749132508 Chr.1: 945677-945677 on Build GRCh38
rs776586302 Chr.1: 945703-945703 on Build GRCh38
rs41285794 Chr.1: 945690-945690 on Build GRCh38
rs372763473 Chr.1: 945659-945659 on Build GRCh38
rs747611760 Chr.1: 945668-945668 on Build GRCh38
rs774830091 Chr.1: 945540-945542 on Build GRCh38
rs760852747 Chr.1: 945598-945601 on Build GRCh38
rs947807398 Chr.1: 945712-945712 on Build GRCh38
rs760834896 Chr.1: 945540-945540 on Build GRCh38
rs936206933 Chr.1: 945691-945691 on Build GRCh38
rs752567395 Chr.1: 945700-945701 on Build GRCh38
rs746155010 Chr.1: 945639-945639 on Build GRCh38
rs759852708 Chr.1: 945614-945614 on Build GRCh38
rs138129954 Chr.1: 945573-945573 on Build GRCh38
rs113650656 Chr.1: 945563-945563 on Build GRCh38
rs760522368 Chr.1: 945635-945635 on Build GRCh38
rs540404478 Chr.1: 945592-945592 on Build GRCh38
rs766192194 Chr.1: 945702-945702 on Build GRCh38
rs187444884 Chr.1: 945542-945542 on Build GRCh38
rs769972766 Chr.1: 945615-945615 on Build GRCh38
rs759589095 Chr.1: 945684-945685 on Build GRCh38
rs781377753 Chr.1: 945611-945611 on Build GRCh38
rs772866895 Chr.1: 945696-945696 on Build GRCh38
rs920336712 Chr.1: 945584-945584 on Build GRCh38
rs748414366 Chr.1: 945584-945585 on Build GRCh38
rs374558436 Chr.1: 945670-945670 on Build GRCh38
rs554710385 Chr.1: 945572-945572 on Build GRCh38
rs777998404 Chr.1: 945667-945667 on Build GRCh38
rs751862886 Chr.1: 945726-945726 on Build GRCh38
rs879100690 Chr.1: 945612-945612 on Build GRCh38
rs772374158 Chr.1: 945585-945599 on Build GRCh38
rs752343648 Chr.1: 945664-945664 on Build GRCh38
rs922598180 Chr.1: 945673-945673 on Build GRCh38
rs369884550 Chr.1: 945537-945537 on Build GRCh38
rs775649472 Chr.1: 945622-945622 on Build GRCh38
rs376096805 Chr.1: 945543-945543 on Build GRCh38
rs747410572 Chr.1: 945608-945608 on Build GRCh38
rs202228302 Chr.1: 945688-945688 on Build GRCh38
rs760712070 Chr.1: 945697-945697 on Build GRCh38
rs781373171 Chr.1: 945674-945674 on Build GRCh38
rs1034323138 Chr.1: 945744-945744 on Build GRCh38
rs758841114 Chr.1: 945665-945665 on Build GRCh38
rs754659814 Chr.1: 945581-945581 on Build GRCh38
rs773244689 Chr.1: 945592-945594 on Build GRCh38
rs983944520 Chr.1: 945620-945620 on Build GRCh38
rs750377234 Chr.1: 945627-945627 on Build GRCh38
rs201671897 Chr.1: 945704-945704 on Build GRCh38
rs765034158 Chr.1: 945683-945684 on Build GRCh38
rs778441416 Chr.1: 945606-945606 on Build GRCh38
rs112433394 Chr.1: 945564-945564 on Build GRCh38
rs112163984 Chr.1: 945562-945562 on Build GRCh38
rs771110900 Chr.1: 945609-945609 on Build GRCh38

View Full Product Details