Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 27577474-27577715 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
KSR1
Entrez Gene ID
8844
Gene Name
kinase suppressor of ras 1
Gene Aliases
KSR, RSU2
UniGene
Hs.133534
Gene Chromosome Location
Chr.17: 27456441-27626438 on Build GRCh38
Transcripts
NM_014238.1 XM_006722154.3 XM_011525429.2 XM_011525430.2 XM_011525431.2 XM_017025267.1 XM_017025268.1 XM_017025269.1 XM_017025270.1 XM_017025271.1 XM_017025272.1 XM_017025273.1 XM_017025274.1 XM_017025275.1 XM_017025276.1 XM_017025279.1 XM_017025280.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
976706 c.460G>A p.G154R Chr.17: 27577579-27577579 on Build GRCh38
976704 c.414G>T p.M138I Chr.17: 27577533-27577533 on Build GRCh38
5510889 c.459C>T p.S153S Chr.17: 27577578-27577578 on Build GRCh38
1609972 c.502C>T p.R168W Chr.17: 27577621-27577621 on Build GRCh38
6263106 c.441G>T p.L147L Chr.17: 27577560-27577560 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs775670428 Chr.17: 27577643-27577643 on Build GRCh38
rs540231409 Chr.17: 27577498-27577498 on Build GRCh38
rs748257885 Chr.17: 27577497-27577497 on Build GRCh38
rs748193495 Chr.17: 27577580-27577580 on Build GRCh38
rs530577825 Chr.17: 27577664-27577664 on Build GRCh38
rs755995795 Chr.17: 27577564-27577564 on Build GRCh38
rs754942098 Chr.17: 27577578-27577578 on Build GRCh38
rs747376747 Chr.17: 27577507-27577507 on Build GRCh38
rs563131445 Chr.17: 27577633-27577633 on Build GRCh38
rs777214460 Chr.17: 27577512-27577512 on Build GRCh38
rs778898995 Chr.17: 27577579-27577579 on Build GRCh38
rs771240772 Chr.17: 27577511-27577511 on Build GRCh38
rs773797374 Chr.17: 27577554-27577554 on Build GRCh38
rs771119796 Chr.17: 27577595-27577595 on Build GRCh38
rs371840328 Chr.17: 27577621-27577621 on Build GRCh38
rs1020754818 Chr.17: 27577527-27577527 on Build GRCh38
rs372194552 Chr.17: 27577591-27577591 on Build GRCh38
rs369843080 Chr.17: 27577680-27577680 on Build GRCh38
rs746238178 Chr.17: 27577607-27577607 on Build GRCh38
rs544947006 Chr.17: 27577622-27577622 on Build GRCh38
rs961543460 Chr.17: 27577572-27577572 on Build GRCh38
rs758402408 Chr.17: 27577588-27577588 on Build GRCh38
rs760009329 Chr.17: 27577540-27577540 on Build GRCh38
rs750058145 Chr.17: 27577562-27577562 on Build GRCh38
rs776900723 Chr.17: 27577599-27577599 on Build GRCh38
rs761371576 Chr.17: 27577556-27577556 on Build GRCh38
rs376492449 Chr.17: 27577677-27577677 on Build GRCh38
rs770260178 Chr.17: 27577614-27577614 on Build GRCh38
rs906283549 Chr.17: 27577533-27577533 on Build GRCh38
rs375680935 Chr.17: 27577570-27577570 on Build GRCh38
rs765744052 Chr.17: 27577547-27577547 on Build GRCh38
rs760448083 Chr.17: 27577627-27577627 on Build GRCh38
rs866740060 Chr.17: 27577499-27577499 on Build GRCh38
rs767284385 Chr.17: 27577557-27577557 on Build GRCh38
rs766327610 Chr.17: 27577565-27577565 on Build GRCh38
rs773362643 Chr.17: 27577502-27577502 on Build GRCh38
rs747180845 Chr.17: 27577594-27577594 on Build GRCh38
rs367832544 Chr.17: 27577616-27577616 on Build GRCh38
rs759558620 Chr.17: 27577574-27577574 on Build GRCh38
rs996366216 Chr.17: 27577567-27577567 on Build GRCh38
rs770477764 Chr.17: 27577686-27577686 on Build GRCh38
rs903853370 Chr.17: 27577583-27577583 on Build GRCh38

View Full Product Details