Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 79406873-79407116 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GK2
Entrez Gene ID
2712
Gene Name
glycerol kinase 2
Gene Aliases
GKP2, GKTA
UniGene
Hs.98008
Gene Chromosome Location
Chr.4: 79406353-79408218 on Build GRCh38
Transcripts
NM_033214.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4409507 c.1206C>G p.F402L Chr.4: 79406995-79406995 on Build GRCh38
7369499 c.1118A>T p.E373V Chr.4: 79407083-79407083 on Build GRCh38
7369497 c.1244G>A p.C415Y Chr.4: 79406957-79406957 on Build GRCh38
1057999 c.1134G>A p.G378G Chr.4: 79407067-79407067 on Build GRCh38
734545 c.1145G>A p.G382D Chr.4: 79407056-79407056 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs749926527 Chr.4: 79406928-79406928 on Build GRCh38
rs765903193 Chr.4: 79407021-79407021 on Build GRCh38
rs750942911 Chr.4: 79407056-79407056 on Build GRCh38
rs1026870089 Chr.4: 79407023-79407023 on Build GRCh38
rs768434357 Chr.4: 79406988-79406988 on Build GRCh38
rs753420892 Chr.4: 79407026-79407026 on Build GRCh38
rs770050343 Chr.4: 79406977-79406977 on Build GRCh38
rs761729595 Chr.4: 79406999-79406999 on Build GRCh38
rs755658600 Chr.4: 79406929-79406929 on Build GRCh38
rs1005241146 Chr.4: 79407084-79407084 on Build GRCh38
rs760126455 Chr.4: 79407020-79407020 on Build GRCh38
rs140956192 Chr.4: 79406942-79406942 on Build GRCh38
rs367978811 Chr.4: 79407008-79407008 on Build GRCh38
rs267600273 Chr.4: 79406943-79406943 on Build GRCh38
rs747491513 Chr.4: 79406948-79406948 on Build GRCh38
rs749086477 Chr.4: 79407047-79407050 on Build GRCh38
rs758932931 Chr.4: 79407074-79407074 on Build GRCh38
rs1015337805 Chr.4: 79407088-79407088 on Build GRCh38
rs779656638 Chr.4: 79406933-79406933 on Build GRCh38
rs774034158 Chr.4: 79406997-79406997 on Build GRCh38
rs988949687 Chr.4: 79407091-79407091 on Build GRCh38
rs777345068 Chr.4: 79406963-79406963 on Build GRCh38
rs113085945 Chr.4: 79406909-79406909 on Build GRCh38
rs771329087 Chr.4: 79406958-79406958 on Build GRCh38
rs769957528 Chr.4: 79407066-79407066 on Build GRCh38
rs145423393 Chr.4: 79406987-79406987 on Build GRCh38
rs866542206 Chr.4: 79407061-79407061 on Build GRCh38
rs747376869 Chr.4: 79407086-79407086 on Build GRCh38
rs140329785 Chr.4: 79406964-79406964 on Build GRCh38
rs746401585 Chr.4: 79407052-79407052 on Build GRCh38
rs200298193 Chr.4: 79407034-79407034 on Build GRCh38
rs965750252 Chr.4: 79407064-79407064 on Build GRCh38
rs757316799 Chr.4: 79407029-79407029 on Build GRCh38
rs867640495 Chr.4: 79406983-79406983 on Build GRCh38
rs778004570 Chr.4: 79406945-79406945 on Build GRCh38
rs142708317 Chr.4: 79406925-79406925 on Build GRCh38
rs1002875280 Chr.4: 79407017-79407017 on Build GRCh38
rs778686178 Chr.4: 79407030-79407030 on Build GRCh38
rs1053783159 Chr.4: 79406934-79406934 on Build GRCh38
rs777466796 Chr.4: 79406987-79406987 on Build GRCh38
rs201409382 Chr.4: 79407038-79407038 on Build GRCh38
rs566437899 Chr.4: 79407011-79407011 on Build GRCh38
rs781759521 Chr.4: 79407048-79407048 on Build GRCh38
rs1033319085 Chr.4: 79407024-79407024 on Build GRCh38

View Full Product Details