Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.20: 33657727-33657998 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
NECAB3
Entrez Gene ID
63941
Gene Name
N-terminal EF-hand calcium binding protein 3
Gene Aliases
APBA2BP, EFCBP3, NIP1, STIP3, SYTIP2, XB51, dJ63M2.4, dJ63M2.5
UniGene
Hs.516986
Gene Chromosome Location
Chr.20: 33657087-33674458 on Build GRCh38
Transcripts
NM_031231.3 NM_031232.3 XM_005260510.1 XM_011528991.1 XM_017028015.1 XM_017028016.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
228731 c.1166C>T p.S389F Chr.20: 33657854-33657854 on Build GRCh38
5484376 c.1127G>A p.R376Q Chr.20: 33657977-33657977 on Build GRCh38
1411258 c.1160delC p.P387fs*6 Chr.20: 33657944-33657944 on Build GRCh38
294610 c.1109T>C p.I370T Chr.20: 33657995-33657995 on Build GRCh38
1197278 c.1121A>G p.H374R Chr.20: 33657983-33657983 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs778569911 Chr.20: 33657979-33657979 on Build GRCh38
rs374364835 Chr.20: 33657862-33657862 on Build GRCh38
rs915413794 Chr.20: 33657948-33657948 on Build GRCh38
rs764436874 Chr.20: 33657911-33657911 on Build GRCh38
rs374672683 Chr.20: 33657801-33657801 on Build GRCh38
rs375312800 Chr.20: 33657809-33657809 on Build GRCh38
rs753159160 Chr.20: 33657879-33657879 on Build GRCh38
rs112148660 Chr.20: 33657941-33657941 on Build GRCh38
rs751747208 Chr.20: 33657931-33657931 on Build GRCh38
rs945606303 Chr.20: 33657949-33657949 on Build GRCh38
rs76653810 Chr.20: 33657843-33657843 on Build GRCh38
rs978659373 Chr.20: 33657957-33657957 on Build GRCh38
rs926602880 Chr.20: 33657887-33657887 on Build GRCh38
rs143415387 Chr.20: 33657853-33657853 on Build GRCh38
rs992404164 Chr.20: 33657923-33657923 on Build GRCh38
rs569505235 Chr.20: 33657823-33657823 on Build GRCh38
rs535208947 Chr.20: 33657934-33657934 on Build GRCh38
rs199832027 Chr.20: 33657939-33657939 on Build GRCh38
rs372436377 Chr.20: 33657935-33657935 on Build GRCh38
rs756094670 Chr.20: 33657963-33657963 on Build GRCh38
rs749146601 Chr.20: 33657977-33657977 on Build GRCh38
rs945628469 Chr.20: 33657753-33657753 on Build GRCh38
rs961201532 Chr.20: 33657818-33657818 on Build GRCh38
rs997898163 Chr.20: 33657802-33657802 on Build GRCh38
rs1049412801 Chr.20: 33657804-33657804 on Build GRCh38
rs1005280625 Chr.20: 33657811-33657811 on Build GRCh38
rs992436833 Chr.20: 33657776-33657776 on Build GRCh38
rs995287129 Chr.20: 33657860-33657860 on Build GRCh38
rs773729552 Chr.20: 33657822-33657822 on Build GRCh38
rs546598082 Chr.20: 33657832-33657832 on Build GRCh38
rs768179067 Chr.20: 33657978-33657978 on Build GRCh38
rs901420983 Chr.20: 33657789-33657789 on Build GRCh38
rs769104107 Chr.20: 33657874-33657874 on Build GRCh38
rs201110040 Chr.20: 33657970-33657970 on Build GRCh38
rs948085672 Chr.20: 33657787-33657787 on Build GRCh38
rs779429439 Chr.20: 33657879-33657888 on Build GRCh38
rs41290872 Chr.20: 33657900-33657900 on Build GRCh38
rs771494532 Chr.20: 33657839-33657839 on Build GRCh38
rs945963142 Chr.20: 33657821-33657821 on Build GRCh38
rs750311270 Chr.20: 33657961-33657961 on Build GRCh38
rs994493300 Chr.20: 33657820-33657820 on Build GRCh38
rs138438360 Chr.20: 33657770-33657770 on Build GRCh38
rs368221322 Chr.20: 33657813-33657813 on Build GRCh38
rs898254871 Chr.20: 33657860-33657860 on Build GRCh38
rs769539565 Chr.20: 33657781-33657781 on Build GRCh38
rs915382520 Chr.20: 33657752-33657752 on Build GRCh38
rs917923336 Chr.20: 33657783-33657783 on Build GRCh38

View Full Product Details