Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 154386236-154386509 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GALNT10
Entrez Gene ID
55568
Gene Name
polypeptide N-acetylgalactosaminyltransferase 10
Gene Aliases
GALNACT10, PPGALNACT10, PPGANTASE10
UniGene
Hs.631797
Gene Chromosome Location
Chr.5: 154190735-154420984 on Build GRCh38
Transcripts
NM_198321.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7319921 c.977G>A p.R326Q Chr.5: 154386351-154386351 on Build GRCh38
1642662 c.1084C>A p.L362I Chr.5: 154386458-154386458 on Build GRCh38
1619881 c.1076G>A p.R359H Chr.5: 154386450-154386450 on Build GRCh38
3919422 c.1015T>C p.L339L Chr.5: 154386389-154386389 on Build GRCh38
5765917 c.1075C>T p.R359C Chr.5: 154386449-154386449 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs189190684 Chr.5: 154386454-154386454 on Build GRCh38
rs144099170 Chr.5: 154386372-154386372 on Build GRCh38
rs202216584 Chr.5: 154386422-154386422 on Build GRCh38
rs374511506 Chr.5: 154386374-154386374 on Build GRCh38
rs771993302 Chr.5: 154386347-154386347 on Build GRCh38
rs146278879 Chr.5: 154386328-154386328 on Build GRCh38
rs756637698 Chr.5: 154386377-154386377 on Build GRCh38
rs554176584 Chr.5: 154386309-154386309 on Build GRCh38
rs572743603 Chr.5: 154386455-154386455 on Build GRCh38
rs778048082 Chr.5: 154386322-154386322 on Build GRCh38
rs751138204 Chr.5: 154386279-154386279 on Build GRCh38
rs773937139 Chr.5: 154386371-154386371 on Build GRCh38
rs752228142 Chr.5: 154386375-154386375 on Build GRCh38
rs764518665 Chr.5: 154386478-154386478 on Build GRCh38
rs937326138 Chr.5: 154386257-154386257 on Build GRCh38
rs201125715 Chr.5: 154386275-154386275 on Build GRCh38
rs146500512 Chr.5: 154386391-154386391 on Build GRCh38
rs768084638 Chr.5: 154386313-154386313 on Build GRCh38
rs139262954 Chr.5: 154386341-154386341 on Build GRCh38
rs1020109208 Chr.5: 154386421-154386421 on Build GRCh38
rs773068024 Chr.5: 154386445-154386445 on Build GRCh38
rs370165932 Chr.5: 154386354-154386354 on Build GRCh38
rs1055727213 Chr.5: 154386274-154386274 on Build GRCh38
rs746044673 Chr.5: 154386405-154386405 on Build GRCh38
rs556767050 Chr.5: 154386383-154386383 on Build GRCh38
rs779298067 Chr.5: 154386394-154386394 on Build GRCh38
rs749095346 Chr.5: 154386449-154386449 on Build GRCh38
rs148741733 Chr.5: 154386425-154386425 on Build GRCh38
rs369551431 Chr.5: 154386277-154386277 on Build GRCh38
rs369609218 Chr.5: 154386266-154386266 on Build GRCh38
rs1019965803 Chr.5: 154386486-154386486 on Build GRCh38
rs952352426 Chr.5: 154386320-154386320 on Build GRCh38
rs758618863 Chr.5: 154386406-154386406 on Build GRCh38
rs757615516 Chr.5: 154386340-154386340 on Build GRCh38
rs376469597 Chr.5: 154386386-154386386 on Build GRCh38
rs377526323 Chr.5: 154386351-154386351 on Build GRCh38
rs756610842 Chr.5: 154386319-154386319 on Build GRCh38
rs764657373 Chr.5: 154386385-154386385 on Build GRCh38
rs1036476132 Chr.5: 154386293-154386293 on Build GRCh38
rs1039768826 Chr.5: 154386408-154386408 on Build GRCh38
rs772327982 Chr.5: 154386296-154386298 on Build GRCh38
rs1014495491 Chr.5: 154386276-154386276 on Build GRCh38
rs921043684 Chr.5: 154386314-154386314 on Build GRCh38
rs760161498 Chr.5: 154386468-154386468 on Build GRCh38
rs185134666 Chr.5: 154386370-154386370 on Build GRCh38
rs759234994 Chr.5: 154386453-154386453 on Build GRCh38
rs776665628 Chr.5: 154386350-154386350 on Build GRCh38
rs1006908887 Chr.5: 154386403-154386403 on Build GRCh38
rs762733735 Chr.5: 154386353-154386353 on Build GRCh38
rs888200737 Chr.5: 154386343-154386343 on Build GRCh38
rs775372012 Chr.5: 154386344-154386344 on Build GRCh38
rs1055201700 Chr.5: 154386373-154386373 on Build GRCh38
rs769690434 Chr.5: 154386439-154386439 on Build GRCh38
rs373356168 Chr.5: 154386301-154386301 on Build GRCh38
rs200481518 Chr.5: 154386450-154386450 on Build GRCh38

View Full Product Details