Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.X: 49304763-49305018 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GAGE10
Entrez Gene ID
102724473
Gene Name
G antigen 10
Gene Aliases
GAGE-10
UniGene
Gene Chromosome Location
Chr.X: 49303646-49319844 on Build GRCh38
Transcripts
NM_001098413.3

SNP Detail

SNP ID Chromosome Location
rs202043957 Chr.X: 49304864-49304864 on Build GRCh38
rs782671769 Chr.X: 49304907-49304907 on Build GRCh38
rs781903889 Chr.X: 49304937-49304937 on Build GRCh38
rs782638456 Chr.X: 49304869-49304869 on Build GRCh38
rs370293504 Chr.X: 49304831-49304831 on Build GRCh38
rs782202314 Chr.X: 49304980-49304980 on Build GRCh38
rs782494383 Chr.X: 49304968-49304968 on Build GRCh38
rs201623415 Chr.X: 49304984-49304984 on Build GRCh38
rs782592082 Chr.X: 49304825-49304825 on Build GRCh38
rs782495276 Chr.X: 49304824-49304824 on Build GRCh38
rs372825163 Chr.X: 49304819-49304819 on Build GRCh38
rs782141108 Chr.X: 49304970-49304971 on Build GRCh38
rs369499720 Chr.X: 49304815-49304815 on Build GRCh38
rs782775961 Chr.X: 49304820-49304820 on Build GRCh38
rs782782853 Chr.X: 49304943-49304943 on Build GRCh38
rs371129235 Chr.X: 49304927-49304927 on Build GRCh38
rs782432918 Chr.X: 49304835-49304835 on Build GRCh38
rs781953021 Chr.X: 49304914-49304914 on Build GRCh38
rs781804380 Chr.X: 49304940-49304940 on Build GRCh38
rs782060702 Chr.X: 49304917-49304917 on Build GRCh38
rs782469432 Chr.X: 49304903-49304903 on Build GRCh38
rs781976782 Chr.X: 49304813-49304813 on Build GRCh38
rs781795393 Chr.X: 49304891-49304891 on Build GRCh38
rs782348477 Chr.X: 49304887-49304887 on Build GRCh38
rs781859648 Chr.X: 49304964-49304964 on Build GRCh38
rs368650707 Chr.X: 49304823-49304823 on Build GRCh38
rs782539541 Chr.X: 49304940-49304940 on Build GRCh38
rs782399323 Chr.X: 49304899-49304899 on Build GRCh38
rs782704814 Chr.X: 49304883-49304884 on Build GRCh38
rs377736170 Chr.X: 49304906-49304906 on Build GRCh38
rs782283385 Chr.X: 49304851-49304851 on Build GRCh38
rs782282321 Chr.X: 49304895-49304895 on Build GRCh38
rs782684277 Chr.X: 49304844-49304844 on Build GRCh38
rs782505969 Chr.X: 49304987-49304988 on Build GRCh38
rs781802977 Chr.X: 49304830-49304830 on Build GRCh38
rs782631403 Chr.X: 49304890-49304890 on Build GRCh38
rs782676385 Chr.X: 49304978-49304978 on Build GRCh38
rs782230048 Chr.X: 49304883-49304883 on Build GRCh38
rs782470925 Chr.X: 49304794-49304794 on Build GRCh38
rs781901073 Chr.X: 49304816-49304816 on Build GRCh38

View Full Product Details