Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 15949516-15949741 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
OR10H4
Entrez Gene ID
126541
Gene Name
olfactory receptor family 10 subfamily H member 4
Gene Aliases
OR19-28
UniGene
Hs.553579
Gene Chromosome Location
Chr.19: 15949008-15949958 on Build GRCh38
Transcripts
NM_001004465.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5056333 c.511G>T p.G171W Chr.19: 15949518-15949518 on Build GRCh38
6588318 c.627G>A p.L209L Chr.19: 15949634-15949634 on Build GRCh38
283497 c.537C>A p.F179L Chr.19: 15949544-15949544 on Build GRCh38
5028511 c.698C>T p.A233V Chr.19: 15949705-15949705 on Build GRCh38
3891797 c.642C>T p.F214F Chr.19: 15949649-15949649 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs748990566 Chr.19: 15949653-15949653 on Build GRCh38
rs771886757 Chr.19: 15949700-15949700 on Build GRCh38
rs751807168 Chr.19: 15949612-15949612 on Build GRCh38
rs753190028 Chr.19: 15949655-15949655 on Build GRCh38
rs768599148 Chr.19: 15949606-15949606 on Build GRCh38
rs778393198 Chr.19: 15949694-15949694 on Build GRCh38
rs114480958 Chr.19: 15949681-15949681 on Build GRCh38
rs758779788 Chr.19: 15949566-15949566 on Build GRCh38
rs138110639 Chr.19: 15949604-15949604 on Build GRCh38
rs115548355 Chr.19: 15949643-15949643 on Build GRCh38
rs367648687 Chr.19: 15949634-15949634 on Build GRCh38
rs887957881 Chr.19: 15949679-15949679 on Build GRCh38
rs747937693 Chr.19: 15949609-15949609 on Build GRCh38
rs147053389 Chr.19: 15949675-15949675 on Build GRCh38
rs753357526 Chr.19: 15949678-15949678 on Build GRCh38
rs755700465 Chr.19: 15949603-15949603 on Build GRCh38
rs897816385 Chr.19: 15949632-15949632 on Build GRCh38
rs370476895 Chr.19: 15949612-15949612 on Build GRCh38
rs61732447 Chr.19: 15949713-15949713 on Build GRCh38
rs565552384 Chr.19: 15949598-15949598 on Build GRCh38
rs147935606 Chr.19: 15949542-15949542 on Build GRCh38
rs150695702 Chr.19: 15949696-15949696 on Build GRCh38
rs763970552 Chr.19: 15949592-15949592 on Build GRCh38
rs375432649 Chr.19: 15949613-15949613 on Build GRCh38
rs141816381 Chr.19: 15949683-15949683 on Build GRCh38
rs764567314 Chr.19: 15949664-15949664 on Build GRCh38
rs563798121 Chr.19: 15949579-15949579 on Build GRCh38
rs745590458 Chr.19: 15949596-15949596 on Build GRCh38
rs751839001 Chr.19: 15949666-15949666 on Build GRCh38
rs577304997 Chr.19: 15949580-15949580 on Build GRCh38
rs370722157 Chr.19: 15949706-15949706 on Build GRCh38
rs546161132 Chr.19: 15949582-15949582 on Build GRCh38
rs777994504 Chr.19: 15949584-15949584 on Build GRCh38
rs759640387 Chr.19: 15949651-15949651 on Build GRCh38
rs753136890 Chr.19: 15949561-15949561 on Build GRCh38
rs779519011 Chr.19: 15949677-15949677 on Build GRCh38
rs899490498 Chr.19: 15949644-15949644 on Build GRCh38
rs765854391 Chr.19: 15949654-15949654 on Build GRCh38
rs777657234 Chr.19: 15949597-15949599 on Build GRCh38
rs146373804 Chr.19: 15949714-15949714 on Build GRCh38
rs929570985 Chr.19: 15949636-15949636 on Build GRCh38
rs761010089 Chr.19: 15949633-15949633 on Build GRCh38
rs140031931 Chr.19: 15949707-15949707 on Build GRCh38
rs868676405 Chr.19: 15949558-15949558 on Build GRCh38

View Full Product Details