Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.20: 21126145-21126372 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
KIZ
Entrez Gene ID
55857
Gene Name
kizuna centrosomal protein
Gene Aliases
C20orf19, HT013, Kizuna, NCRNA00153, PLK1S1, RP69
UniGene
Hs.187635
Gene Chromosome Location
Chr.20: 21125980-21246628 on Build GRCh38
Transcripts
NM_001163022.1 NM_001163023.1 NM_001276389.1 NM_018474.4 XM_011529296.2 XM_011529297.2 XM_011529298.2 XM_011529299.2 XM_017027951.1 XM_017027952.1 XM_017027953.1 XM_017027954.1

SNP Detail

SNP ID Chromosome Location
rs377745565 Chr.20: 21126284-21126284 on Build GRCh38
rs554276789 Chr.20: 21126264-21126264 on Build GRCh38
rs541555909 Chr.20: 21126210-21126210 on Build GRCh38
rs764064292 Chr.20: 21126220-21126220 on Build GRCh38
rs1016600203 Chr.20: 21126324-21126324 on Build GRCh38
rs775312973 Chr.20: 21126165-21126165 on Build GRCh38
rs1035962282 Chr.20: 21126338-21126338 on Build GRCh38
rs913715921 Chr.20: 21126180-21126180 on Build GRCh38
rs750974457 Chr.20: 21126265-21126265 on Build GRCh38
rs921636473 Chr.20: 21126190-21126190 on Build GRCh38
rs970067842 Chr.20: 21126312-21126312 on Build GRCh38
rs887945850 Chr.20: 21126304-21126304 on Build GRCh38
rs1029316934 Chr.20: 21126345-21126345 on Build GRCh38
rs578218964 Chr.20: 21126258-21126258 on Build GRCh38
rs996439698 Chr.20: 21126341-21126341 on Build GRCh38
rs531308909 Chr.20: 21126303-21126303 on Build GRCh38
rs547855068 Chr.20: 21126315-21126315 on Build GRCh38
rs899527190 Chr.20: 21126332-21126332 on Build GRCh38
rs911905109 Chr.20: 21126226-21126226 on Build GRCh38
rs891870467 Chr.20: 21126289-21126289 on Build GRCh38
rs988408197 Chr.20: 21126179-21126179 on Build GRCh38
rs931877776 Chr.20: 21126230-21126230 on Build GRCh38
rs138913800 Chr.20: 21126235-21126235 on Build GRCh38
rs868175109 Chr.20: 21126311-21126311 on Build GRCh38
rs1021580500 Chr.20: 21126293-21126293 on Build GRCh38
rs587777376 Chr.20: 21126167-21126167 on Build GRCh38
rs115300323 Chr.20: 21126291-21126291 on Build GRCh38
rs890247776 Chr.20: 21126246-21126246 on Build GRCh38
rs929558763 Chr.20: 21126285-21126285 on Build GRCh38
rs761910040 Chr.20: 21126242-21126242 on Build GRCh38

View Full Product Details