Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 40363667-40363918 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GJD3
Entrez Gene ID
125111
Gene Name
gap junction protein delta 3
Gene Aliases
CX31.9, Cx30.2, GJA11, GJC1
UniGene
Hs.444663
Gene Chromosome Location
Chr.17: 40360652-40364693 on Build GRCh38
Transcripts
NM_152219.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7106052 c.142G>A p.E48K Chr.17: 40363674-40363674 on Build GRCh38
4066173 c.56C>T p.P19L Chr.17: 40363760-40363760 on Build GRCh38
5740973 c.138_140delGGA p.E46delE Chr.17: 40363676-40363678 on Build GRCh38
7447994 c.110T>C p.L37P Chr.17: 40363706-40363706 on Build GRCh38
5037360 c.51G>C p.Q17H Chr.17: 40363765-40363765 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs753783392 Chr.17: 40363850-40363850 on Build GRCh38
rs377412692 Chr.17: 40363832-40363832 on Build GRCh38
rs942297138 Chr.17: 40363762-40363762 on Build GRCh38
rs993928072 Chr.17: 40363851-40363851 on Build GRCh38
rs774352443 Chr.17: 40363763-40363763 on Build GRCh38
rs752963630 Chr.17: 40363697-40363697 on Build GRCh38
rs933447288 Chr.17: 40363691-40363691 on Build GRCh38
rs376380005 Chr.17: 40363755-40363755 on Build GRCh38
rs779484098 Chr.17: 40363846-40363846 on Build GRCh38
rs747977244 Chr.17: 40363868-40363868 on Build GRCh38
rs950036353 Chr.17: 40363774-40363774 on Build GRCh38
rs561136184 Chr.17: 40363852-40363852 on Build GRCh38
rs930743765 Chr.17: 40363715-40363715 on Build GRCh38
rs745400614 Chr.17: 40363750-40363750 on Build GRCh38
rs751927921 Chr.17: 40363805-40363805 on Build GRCh38
rs373332316 Chr.17: 40363712-40363712 on Build GRCh38
rs774084905 Chr.17: 40363809-40363809 on Build GRCh38
rs770583005 Chr.17: 40363780-40363780 on Build GRCh38
rs753810053 Chr.17: 40363776-40363776 on Build GRCh38
rs1045988974 Chr.17: 40363775-40363775 on Build GRCh38
rs534231740 Chr.17: 40363866-40363866 on Build GRCh38
rs373039837 Chr.17: 40363756-40363756 on Build GRCh38
rs922366098 Chr.17: 40363686-40363686 on Build GRCh38
rs1050592450 Chr.17: 40363736-40363736 on Build GRCh38
rs190167816 Chr.17: 40363804-40363804 on Build GRCh38
rs757706657 Chr.17: 40363811-40363811 on Build GRCh38
rs893333176 Chr.17: 40363873-40363873 on Build GRCh38
rs767574471 Chr.17: 40363769-40363769 on Build GRCh38
rs745333134 Chr.17: 40363825-40363825 on Build GRCh38
rs771862649 Chr.17: 40363893-40363893 on Build GRCh38
rs1019999010 Chr.17: 40363896-40363896 on Build GRCh38
rs542921075 Chr.17: 40363819-40363819 on Build GRCh38
rs376249337 Chr.17: 40363830-40363830 on Build GRCh38
rs748940271 Chr.17: 40363848-40363848 on Build GRCh38
rs763807233 Chr.17: 40363752-40363753 on Build GRCh38
rs560180899 Chr.17: 40363782-40363782 on Build GRCh38
rs771496279 Chr.17: 40363729-40363729 on Build GRCh38
rs780504404 Chr.17: 40363744-40363744 on Build GRCh38
rs906124847 Chr.17: 40363838-40363838 on Build GRCh38
rs758678981 Chr.17: 40363802-40363802 on Build GRCh38
rs747502161 Chr.17: 40363719-40363719 on Build GRCh38
rs1009980401 Chr.17: 40363891-40363891 on Build GRCh38
rs761773257 Chr.17: 40363768-40363768 on Build GRCh38
rs757150775 Chr.17: 40363861-40363868 on Build GRCh38
rs759691964 Chr.17: 40363777-40363777 on Build GRCh38
rs758730521 Chr.17: 40363699-40363699 on Build GRCh38

View Full Product Details