Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 99173597-99173838 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
KPNA7
Entrez Gene ID
402569
Gene Name
karyopherin subunit alpha 7
Gene Aliases
IPOA8
UniGene
Hs.253050
Gene Chromosome Location
Chr.7: 99145467-99219417 on Build GRCh38
Transcripts
XM_011516215.2 XM_017012211.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5062710 c.1548_1549insA p.*517fs? Chr.7: 99173710-99173711 on Build GRCh38
5884615 c.1479C>G p.S493R Chr.7: 99173780-99173780 on Build GRCh38
4759213 c.1472A>G p.D491G Chr.7: 99173787-99173787 on Build GRCh38
1623367 c.1479C>T p.S493S Chr.7: 99173780-99173780 on Build GRCh38
3431928 c.1550A>G p.*517W Chr.7: 99173709-99173709 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs759978557 Chr.7: 99173805-99173805 on Build GRCh38
rs976179206 Chr.7: 99173777-99173777 on Build GRCh38
rs761158968 Chr.7: 99173800-99173800 on Build GRCh38
rs1006434442 Chr.7: 99173685-99173685 on Build GRCh38
rs148988263 Chr.7: 99173786-99173786 on Build GRCh38
rs976134179 Chr.7: 99173658-99173658 on Build GRCh38
rs371763807 Chr.7: 99173736-99173736 on Build GRCh38
rs537532950 Chr.7: 99173729-99173729 on Build GRCh38
rs554796491 Chr.7: 99173703-99173703 on Build GRCh38
rs367591876 Chr.7: 99173728-99173728 on Build GRCh38
rs767922702 Chr.7: 99173754-99173754 on Build GRCh38
rs964768470 Chr.7: 99173772-99173772 on Build GRCh38
rs967097269 Chr.7: 99173808-99173808 on Build GRCh38
rs887734258 Chr.7: 99173647-99173647 on Build GRCh38
rs1026648371 Chr.7: 99173646-99173646 on Build GRCh38
rs570210924 Chr.7: 99173726-99173726 on Build GRCh38
rs745830416 Chr.7: 99173665-99173665 on Build GRCh38
rs774947420 Chr.7: 99173752-99173752 on Build GRCh38
rs374674531 Chr.7: 99173813-99173813 on Build GRCh38
rs771798458 Chr.7: 99173678-99173678 on Build GRCh38
rs374639225 Chr.7: 99173753-99173753 on Build GRCh38
rs868137944 Chr.7: 99173743-99173743 on Build GRCh38
rs145752831 Chr.7: 99173751-99173751 on Build GRCh38

View Full Product Details