Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 222671647-222671872 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
MOGAT1
Entrez Gene ID
116255
Gene Name
monoacylglycerol O-acyltransferase 1
Gene Aliases
DGAT2L, DGAT2L1, MGAT1
UniGene
Hs.344090
Gene Chromosome Location
Chr.2: 222671738-222709930 on Build GRCh38
Transcripts
NM_058165.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3953038 c.83A>G p.Y28C Chr.2: 222671868-222671868 on Build GRCh38
5396332 c.72G>A p.W24* Chr.2: 222671857-222671857 on Build GRCh38
6274925 c.66G>T p.L22L Chr.2: 222671851-222671851 on Build GRCh38
5396331 c.72G>A p.W24* Chr.2: 222671857-222671857 on Build GRCh38
6274924 c.66G>T p.L22L Chr.2: 222671851-222671851 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs892648427 Chr.2: 222671671-222671671 on Build GRCh38
rs754187116 Chr.2: 222671787-222671787 on Build GRCh38
rs971830359 Chr.2: 222671783-222671783 on Build GRCh38
rs575399876 Chr.2: 222671845-222671845 on Build GRCh38
rs35959734 Chr.2: 222671822-222671822 on Build GRCh38
rs760992318 Chr.2: 222671763-222671763 on Build GRCh38
rs535627830 Chr.2: 222671741-222671741 on Build GRCh38
rs779344608 Chr.2: 222671813-222671813 on Build GRCh38
rs566599483 Chr.2: 222671738-222671738 on Build GRCh38
rs777274652 Chr.2: 222671831-222671831 on Build GRCh38
rs1019080270 Chr.2: 222671759-222671759 on Build GRCh38
rs188664940 Chr.2: 222671690-222671690 on Build GRCh38
rs1022557086 Chr.2: 222671695-222671695 on Build GRCh38
rs1041723364 Chr.2: 222671668-222671668 on Build GRCh38
rs750607894 Chr.2: 222671747-222671747 on Build GRCh38
rs766718950 Chr.2: 222671768-222671768 on Build GRCh38
rs904147527 Chr.2: 222671705-222671705 on Build GRCh38
rs924044100 Chr.2: 222671801-222671823 on Build GRCh38
rs546713980 Chr.2: 222671734-222671734 on Build GRCh38
rs1028688600 Chr.2: 222671715-222671715 on Build GRCh38
rs755487022 Chr.2: 222671798-222671798 on Build GRCh38
rs570308864 Chr.2: 222671696-222671696 on Build GRCh38
rs920425256 Chr.2: 222671792-222671792 on Build GRCh38
rs939320727 Chr.2: 222671850-222671850 on Build GRCh38
rs370650786 Chr.2: 222671828-222671828 on Build GRCh38
rs41272717 Chr.2: 222671709-222671709 on Build GRCh38
rs961909302 Chr.2: 222671766-222671766 on Build GRCh38
rs1050614183 Chr.2: 222671739-222671739 on Build GRCh38

View Full Product Details