Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 49599399-49599666 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FAM186B
Entrez Gene ID
84070
Gene Name
family with sequence similarity 186 member B
Gene Aliases
C12orf25
UniGene
Hs.524406
Gene Chromosome Location
Chr.12: 49582875-49605710 on Build GRCh38
Transcripts
NM_032130.2 NR_027450.1 XM_006719625.2 XM_006719626.2 XM_006719627.3 XM_011538796.2 XM_011538797.2 XM_017020008.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4946215 c.1795C>T p.R599C Chr.12: 49599575-49599575 on Build GRCh38
4682791 c.1840G>A p.A614T Chr.12: 49599530-49599530 on Build GRCh38
1606220 c.2058C>T p.H686H Chr.12: 49599582-49599582 on Build GRCh38
3416898 c.2065C>T p.R689C Chr.12: 49599575-49599575 on Build GRCh38
6867051 c.1778G>A p.R593K Chr.12: 49599592-49599592 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs997948557 Chr.12: 49599505-49599505 on Build GRCh38
rs748591097 Chr.12: 49599631-49599631 on Build GRCh38
rs376783093 Chr.12: 49599575-49599575 on Build GRCh38
rs781163043 Chr.12: 49599517-49599517 on Build GRCh38
rs142724836 Chr.12: 49599611-49599611 on Build GRCh38
rs767793359 Chr.12: 49599458-49599458 on Build GRCh38
rs749541895 Chr.12: 49599623-49599623 on Build GRCh38
rs367804678 Chr.12: 49599425-49599425 on Build GRCh38
rs749905477 Chr.12: 49599469-49599469 on Build GRCh38
rs748303576 Chr.12: 49599542-49599544 on Build GRCh38
rs770017743 Chr.12: 49599541-49599542 on Build GRCh38
rs754744243 Chr.12: 49599467-49599475 on Build GRCh38
rs762255365 Chr.12: 49599543-49599543 on Build GRCh38
rs755208567 Chr.12: 49599624-49599624 on Build GRCh38
rs779887029 Chr.12: 49599432-49599433 on Build GRCh38
rs774806166 Chr.12: 49599542-49599542 on Build GRCh38
rs141058026 Chr.12: 49599629-49599629 on Build GRCh38
rs752498228 Chr.12: 49599599-49599599 on Build GRCh38
rs533213469 Chr.12: 49599574-49599574 on Build GRCh38
rs753480309 Chr.12: 49599481-49599481 on Build GRCh38
rs781034718 Chr.12: 49599503-49599504 on Build GRCh38
rs201727387 Chr.12: 49599470-49599470 on Build GRCh38
rs199689151 Chr.12: 49599461-49599461 on Build GRCh38
rs375483333 Chr.12: 49599484-49599484 on Build GRCh38
rs1051178056 Chr.12: 49599520-49599520 on Build GRCh38
rs769154684 Chr.12: 49599538-49599538 on Build GRCh38
rs572186499 Chr.12: 49599530-49599530 on Build GRCh38
rs752950919 Chr.12: 49599531-49599531 on Build GRCh38
rs746000838 Chr.12: 49599529-49599529 on Build GRCh38
rs751553992 Chr.12: 49599448-49599448 on Build GRCh38
rs969951410 Chr.12: 49599551-49599551 on Build GRCh38
rs766064665 Chr.12: 49599474-49599474 on Build GRCh38
rs753496585 Chr.12: 49599573-49599573 on Build GRCh38
rs1008763739 Chr.12: 49599432-49599432 on Build GRCh38
rs960749594 Chr.12: 49599579-49599579 on Build GRCh38
rs143957189 Chr.12: 49599555-49599555 on Build GRCh38
rs201920092 Chr.12: 49599485-49599485 on Build GRCh38
rs758309157 Chr.12: 49599495-49599495 on Build GRCh38
rs372195064 Chr.12: 49599533-49599533 on Build GRCh38
rs764142123 Chr.12: 49599608-49599608 on Build GRCh38
rs758098194 Chr.12: 49599606-49599606 on Build GRCh38
rs773566147 Chr.12: 49599553-49599553 on Build GRCh38
rs148217891 Chr.12: 49599560-49599560 on Build GRCh38
rs147347392 Chr.12: 49599617-49599617 on Build GRCh38
rs980425603 Chr.12: 49599618-49599618 on Build GRCh38
rs765872651 Chr.12: 49599571-49599571 on Build GRCh38
rs756266654 Chr.12: 49599612-49599612 on Build GRCh38
rs1044341888 Chr.12: 49599455-49599455 on Build GRCh38
rs772671883 Chr.12: 49599546-49599546 on Build GRCh38

View Full Product Details