Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 41737603-41737866 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PGC
Entrez Gene ID
5225
Gene Name
progastricsin
Gene Aliases
PEPC, PGII
UniGene
Hs.1867
Gene Chromosome Location
Chr.6: 41736711-41747401 on Build GRCh38
Transcripts
NM_002630.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4713791 c.946C>A p.L316M Chr.6: 41737798-41737798 on Build GRCh38
3078708 c.1001C>G p.S334C Chr.6: 41737743-41737743 on Build GRCh38
6744589 c.989T>C p.L330P Chr.6: 41737755-41737755 on Build GRCh38
5953287 c.921C>A p.L307L Chr.6: 41737823-41737823 on Build GRCh38
3873994 c.991C>G p.P331A Chr.6: 41737753-41737753 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs779227115 Chr.6: 41737752-41737752 on Build GRCh38
rs760974781 Chr.6: 41737835-41737835 on Build GRCh38
rs973844500 Chr.6: 41737678-41737678 on Build GRCh38
rs770753897 Chr.6: 41737804-41737805 on Build GRCh38
rs201949514 Chr.6: 41737709-41737709 on Build GRCh38
rs144666615 Chr.6: 41737715-41737715 on Build GRCh38
rs267601031 Chr.6: 41737759-41737759 on Build GRCh38
rs201999322 Chr.6: 41737661-41737665 on Build GRCh38
rs988065079 Chr.6: 41737749-41737749 on Build GRCh38
rs759631380 Chr.6: 41737806-41737807 on Build GRCh38
rs573861763 Chr.6: 41737754-41737754 on Build GRCh38
rs963858698 Chr.6: 41737658-41737658 on Build GRCh38
rs142306545 Chr.6: 41737812-41737812 on Build GRCh38
rs776649957 Chr.6: 41737807-41737807 on Build GRCh38
rs775030351 Chr.6: 41737820-41737820 on Build GRCh38
rs776891566 Chr.6: 41737805-41737806 on Build GRCh38
rs746804878 Chr.6: 41737803-41737804 on Build GRCh38
rs201287149 Chr.6: 41737773-41737773 on Build GRCh38
rs112599241 Chr.6: 41737720-41737720 on Build GRCh38
rs770994953 Chr.6: 41737793-41737793 on Build GRCh38
rs200403445 Chr.6: 41737813-41737813 on Build GRCh38
rs775368337 Chr.6: 41737807-41737808 on Build GRCh38
rs754966619 Chr.6: 41737751-41737751 on Build GRCh38
rs116237306 Chr.6: 41737657-41737657 on Build GRCh38
rs750794307 Chr.6: 41737832-41737832 on Build GRCh38
rs753252461 Chr.6: 41737804-41737804 on Build GRCh38
rs753967186 Chr.6: 41737731-41737731 on Build GRCh38
rs537818153 Chr.6: 41737655-41737655 on Build GRCh38
rs369120707 Chr.6: 41737775-41737775 on Build GRCh38
rs763142187 Chr.6: 41737831-41737832 on Build GRCh38
rs746793953 Chr.6: 41737792-41737792 on Build GRCh38
rs764954045 Chr.6: 41737689-41737689 on Build GRCh38
rs774401575 Chr.6: 41737822-41737822 on Build GRCh38
rs1034946683 Chr.6: 41737630-41737630 on Build GRCh38
rs762266223 Chr.6: 41737699-41737699 on Build GRCh38

View Full Product Details