Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 144109739-144109990 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
OR2A2
Entrez Gene ID
442361
Gene Name
olfactory receptor family 2 subfamily A member 2
Gene Aliases
OR2A17P, OR2A2P, OR7-11, OST008
UniGene
Hs.553841
Gene Chromosome Location
Chr.7: 144109583-144110539 on Build GRCh38
Transcripts
NM_001005480.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4941042 c.366T>A p.Y122* Chr.7: 144109948-144109948 on Build GRCh38
4996805 c.254T>C p.M85T Chr.7: 144109836-144109836 on Build GRCh38
262572 c.297G>T p.Q99H Chr.7: 144109879-144109879 on Build GRCh38
246218 c.165C>T p.H55H Chr.7: 144109747-144109747 on Build GRCh38
3781278 c.259C>A p.Q87K Chr.7: 144109841-144109841 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs759170009 Chr.7: 144109768-144109768 on Build GRCh38
rs750337811 Chr.7: 144109947-144109947 on Build GRCh38
rs371679975 Chr.7: 144109865-144109865 on Build GRCh38
rs959961776 Chr.7: 144109945-144109945 on Build GRCh38
rs201869620 Chr.7: 144109948-144109948 on Build GRCh38
rs184278314 Chr.7: 144109967-144109967 on Build GRCh38
rs752503274 Chr.7: 144109795-144109795 on Build GRCh38
rs758153376 Chr.7: 144109797-144109797 on Build GRCh38
rs762496494 Chr.7: 144109931-144109931 on Build GRCh38
rs780091757 Chr.7: 144109825-144109825 on Build GRCh38
rs2684427 Chr.7: 144109831-144109831 on Build GRCh38
rs770965586 Chr.7: 144109820-144109820 on Build GRCh38
rs767179454 Chr.7: 144109898-144109898 on Build GRCh38
rs757082690 Chr.7: 144109875-144109875 on Build GRCh38
rs948539474 Chr.7: 144109819-144109819 on Build GRCh38
rs759151032 Chr.7: 144109856-144109856 on Build GRCh38
rs760172536 Chr.7: 144109917-144109917 on Build GRCh38
rs1020215589 Chr.7: 144109866-144109866 on Build GRCh38
rs775778361 Chr.7: 144109956-144109957 on Build GRCh38
rs746887137 Chr.7: 144109812-144109812 on Build GRCh38
rs367713543 Chr.7: 144109888-144109888 on Build GRCh38
rs770169735 Chr.7: 144109882-144109882 on Build GRCh38
rs61734798 Chr.7: 144109841-144109841 on Build GRCh38
rs761107857 Chr.7: 144109896-144109896 on Build GRCh38
rs61734817 Chr.7: 144109884-144109884 on Build GRCh38
rs777422624 Chr.7: 144109871-144109871 on Build GRCh38
rs756111052 Chr.7: 144109949-144109949 on Build GRCh38
rs61734829 Chr.7: 144109915-144109915 on Build GRCh38
rs915775392 Chr.7: 144109817-144109817 on Build GRCh38
rs61734823 Chr.7: 144109905-144109905 on Build GRCh38
rs764804289 Chr.7: 144109921-144109921 on Build GRCh38
rs144874807 Chr.7: 144109847-144109847 on Build GRCh38
rs778715655 Chr.7: 144109789-144109789 on Build GRCh38
rs753459210 Chr.7: 144109770-144109770 on Build GRCh38
rs375499730 Chr.7: 144109942-144109942 on Build GRCh38
rs200096149 Chr.7: 144109890-144109890 on Build GRCh38
rs763818273 Chr.7: 144109940-144109940 on Build GRCh38
rs375475646 Chr.7: 144109891-144109891 on Build GRCh38
rs142627182 Chr.7: 144109784-144109784 on Build GRCh38
rs139387901 Chr.7: 144109848-144109848 on Build GRCh38
rs778951259 Chr.7: 144109951-144109951 on Build GRCh38
rs61734821 Chr.7: 144109895-144109895 on Build GRCh38
rs781110473 Chr.7: 144109882-144109882 on Build GRCh38
rs574785094 Chr.7: 144109836-144109836 on Build GRCh38
rs191243519 Chr.7: 144109854-144109854 on Build GRCh38
rs771526497 Chr.7: 144109902-144109902 on Build GRCh38
rs761285796 Chr.7: 144109766-144109766 on Build GRCh38
rs749324500 Chr.7: 144109885-144109885 on Build GRCh38
rs2684428 Chr.7: 144109834-144109834 on Build GRCh38
rs374195775 Chr.7: 144109840-144109840 on Build GRCh38
rs978122275 Chr.7: 144109877-144109877 on Build GRCh38
rs544154038 Chr.7: 144109764-144109764 on Build GRCh38
rs61734820 Chr.7: 144109894-144109894 on Build GRCh38
rs200492121 Chr.7: 144109832-144109832 on Build GRCh38
rs758105031 Chr.7: 144109870-144109870 on Build GRCh38
rs768744521 Chr.7: 144109886-144109886 on Build GRCh38
rs372540292 Chr.7: 144109927-144109927 on Build GRCh38
rs902782083 Chr.7: 144109920-144109920 on Build GRCh38
rs767392385 Chr.7: 144109946-144109946 on Build GRCh38
rs374526958 Chr.7: 144109873-144109873 on Build GRCh38
rs371015719 Chr.7: 144109810-144109810 on Build GRCh38
rs757105007 Chr.7: 144109944-144109944 on Build GRCh38
rs61729718 Chr.7: 144109862-144109862 on Build GRCh38
rs1031257932 Chr.7: 144109775-144109775 on Build GRCh38

View Full Product Details