Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 235990945-235991194 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
NID1
Entrez Gene ID
4811
Gene Name
nidogen 1
Gene Aliases
NID
UniGene
Hs.356624
Gene Chromosome Location
Chr.1: 235975830-236065181 on Build GRCh38
Transcripts
NM_002508.2 XM_011544195.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6125275 c.2832C>A p.T944T Chr.1: 235990982-235990982 on Build GRCh38
1748241 c.2796G>T p.A932A Chr.1: 235991018-235991018 on Build GRCh38
905951 c.2866C>T p.R956C Chr.1: 235990948-235990948 on Build GRCh38
5425211 c.2756G>T p.C919F Chr.1: 235991058-235991058 on Build GRCh38
246034 c.2809G>A p.V937M Chr.1: 235991005-235991005 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs776442794 Chr.1: 235991030-235991030 on Build GRCh38
rs557330114 Chr.1: 235991009-235991009 on Build GRCh38
rs375973144 Chr.1: 235991036-235991036 on Build GRCh38
rs780745362 Chr.1: 235990992-235990992 on Build GRCh38
rs35271803 Chr.1: 235991016-235991017 on Build GRCh38
rs749275342 Chr.1: 235991104-235991104 on Build GRCh38
rs3738528 Chr.1: 235991129-235991129 on Build GRCh38
rs765322640 Chr.1: 235990968-235990968 on Build GRCh38
rs11404777 Chr.1: 235991116-235991117 on Build GRCh38
rs767951235 Chr.1: 235991063-235991063 on Build GRCh38
rs559829932 Chr.1: 235991140-235991140 on Build GRCh38
rs755447617 Chr.1: 235991004-235991004 on Build GRCh38
rs569425269 Chr.1: 235991128-235991129 on Build GRCh38
rs573754065 Chr.1: 235991091-235991091 on Build GRCh38
rs979644250 Chr.1: 235991035-235991035 on Build GRCh38
rs745412828 Chr.1: 235991015-235991015 on Build GRCh38
rs200467845 Chr.1: 235991005-235991005 on Build GRCh38
rs763009822 Chr.1: 235991039-235991039 on Build GRCh38
rs67633802 Chr.1: 235991129-235991129 on Build GRCh38
rs796719314 Chr.1: 235991128-235991128 on Build GRCh38
rs4660137 Chr.1: 235991065-235991065 on Build GRCh38
rs200352517 Chr.1: 235990979-235990979 on Build GRCh38
rs751967777 Chr.1: 235991000-235991000 on Build GRCh38
rs1041310798 Chr.1: 235991088-235991088 on Build GRCh38
rs148296373 Chr.1: 235990985-235990985 on Build GRCh38
rs371585180 Chr.1: 235991087-235991087 on Build GRCh38
rs540843863 Chr.1: 235991138-235991138 on Build GRCh38
rs199783074 Chr.1: 235991062-235991062 on Build GRCh38
rs536964808 Chr.1: 235991129-235991130 on Build GRCh38
rs1046899846 Chr.1: 235991133-235991133 on Build GRCh38
rs370883577 Chr.1: 235991130-235991131 on Build GRCh38
rs768374026 Chr.1: 235991029-235991029 on Build GRCh38
rs757845906 Chr.1: 235991092-235991092 on Build GRCh38
rs533454229 Chr.1: 235991144-235991144 on Build GRCh38
rs138322087 Chr.1: 235991019-235991019 on Build GRCh38
rs779545735 Chr.1: 235991099-235991099 on Build GRCh38
rs796784845 Chr.1: 235991116-235991117 on Build GRCh38
rs147031904 Chr.1: 235991008-235991008 on Build GRCh38
rs992082328 Chr.1: 235991040-235991040 on Build GRCh38
rs771027111 Chr.1: 235991117-235991118 on Build GRCh38
rs3831991 Chr.1: 235991115-235991116 on Build GRCh38
rs150844582 Chr.1: 235991006-235991006 on Build GRCh38
rs752051457 Chr.1: 235991059-235991059 on Build GRCh38
rs762044163 Chr.1: 235991034-235991034 on Build GRCh38
rs370999601 Chr.1: 235991018-235991018 on Build GRCh38
rs756906039 Chr.1: 235991067-235991067 on Build GRCh38
rs917958750 Chr.1: 235991061-235991061 on Build GRCh38
rs146230129 Chr.1: 235991023-235991023 on Build GRCh38
rs373483665 Chr.1: 235990990-235990990 on Build GRCh38
rs766975681 Chr.1: 235991053-235991053 on Build GRCh38
rs946452738 Chr.1: 235991044-235991044 on Build GRCh38
rs996990612 Chr.1: 235991128-235991129 on Build GRCh38
rs1016074198 Chr.1: 235991166-235991166 on Build GRCh38

View Full Product Details