Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 58487575-58487814 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
HSF5
Entrez Gene ID
124535
Gene Name
heat shock transcription factor 5
Gene Aliases
HSF 5, HSTF 5
UniGene
Hs.380061
Gene Chromosome Location
Chr.17: 58420167-58488401 on Build GRCh38
Transcripts
NM_001080439.2 XM_011524283.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
706676 c.528G>C p.A176A Chr.17: 58487747-58487747 on Build GRCh38
6147076 c.478G>A p.A160T Chr.17: 58487797-58487797 on Build GRCh38
6037981 c.518C>A p.P173Q Chr.17: 58487757-58487757 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs151255293 Chr.17: 58487664-58487665 on Build GRCh38
rs892947938 Chr.17: 58487755-58487755 on Build GRCh38
rs754983469 Chr.17: 58487721-58487721 on Build GRCh38
rs1051962613 Chr.17: 58487609-58487609 on Build GRCh38
rs889334954 Chr.17: 58487619-58487619 on Build GRCh38
rs761030096 Chr.17: 58487699-58487699 on Build GRCh38
rs974231840 Chr.17: 58487645-58487645 on Build GRCh38
rs774435362 Chr.17: 58487777-58487777 on Build GRCh38
rs936747961 Chr.17: 58487744-58487744 on Build GRCh38
rs543934308 Chr.17: 58487764-58487764 on Build GRCh38
rs978154011 Chr.17: 58487732-58487732 on Build GRCh38
rs1007531032 Chr.17: 58487628-58487628 on Build GRCh38
rs912579407 Chr.17: 58487709-58487709 on Build GRCh38
rs906850725 Chr.17: 58487601-58487601 on Build GRCh38
rs764357665 Chr.17: 58487706-58487706 on Build GRCh38
rs539787620 Chr.17: 58487607-58487607 on Build GRCh38
rs771099616 Chr.17: 58487768-58487768 on Build GRCh38
rs558293095 Chr.17: 58487611-58487611 on Build GRCh38
rs759553917 Chr.17: 58487778-58487778 on Build GRCh38
rs554423594 Chr.17: 58487599-58487599 on Build GRCh38
rs776128232 Chr.17: 58487787-58487787 on Build GRCh38
rs1027567355 Chr.17: 58487676-58487676 on Build GRCh38
rs751068231 Chr.17: 58487717-58487717 on Build GRCh38
rs576559569 Chr.17: 58487734-58487734 on Build GRCh38
rs779745036 Chr.17: 58487712-58487712 on Build GRCh38
rs954346654 Chr.17: 58487682-58487682 on Build GRCh38
rs1021559301 Chr.17: 58487641-58487641 on Build GRCh38
rs769689205 Chr.17: 58487742-58487742 on Build GRCh38
rs572372490 Chr.17: 58487600-58487600 on Build GRCh38
rs777398359 Chr.17: 58487751-58487753 on Build GRCh38
rs199938494 Chr.17: 58487782-58487782 on Build GRCh38
rs781213617 Chr.17: 58487729-58487729 on Build GRCh38
rs757963167 Chr.17: 58487711-58487711 on Build GRCh38
rs1002643153 Chr.17: 58487620-58487620 on Build GRCh38
rs772572277 Chr.17: 58487779-58487779 on Build GRCh38
rs961547070 Chr.17: 58487640-58487640 on Build GRCh38
rs750144596 Chr.17: 58487707-58487707 on Build GRCh38
rs749565745 Chr.17: 58487765-58487765 on Build GRCh38
rs746140423 Chr.17: 58487750-58487751 on Build GRCh38
rs111702087 Chr.17: 58487718-58487718 on Build GRCh38

View Full Product Details