Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 53967564-53967833 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
LRRC42
Entrez Gene ID
115353
Gene Name
leucine rich repeat containing 42
Gene Aliases
dJ167A19.4
UniGene
Hs.40094
Gene Chromosome Location
Chr.1: 53946077-53968168 on Build GRCh38
Transcripts
NM_001256409.1 NM_052940.4 XM_006710327.3 XM_006710328.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5015831 c.1028G>A p.R343Q Chr.1: 53967680-53967680 on Build GRCh38
910825 c.1095C>A p.F365L Chr.1: 53967747-53967747 on Build GRCh38
464771 c.1088T>A p.L363H Chr.1: 53967740-53967740 on Build GRCh38
6572051 c.1033G>C p.E345Q Chr.1: 53967685-53967685 on Build GRCh38
1602549 c.1041A>G p.P347P Chr.1: 53967693-53967693 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs192607357 Chr.1: 53967672-53967672 on Build GRCh38
rs150193165 Chr.1: 53967781-53967781 on Build GRCh38
rs201664743 Chr.1: 53967649-53967649 on Build GRCh38
rs747838150 Chr.1: 53967777-53967777 on Build GRCh38
rs747493961 Chr.1: 53967731-53967731 on Build GRCh38
rs145405790 Chr.1: 53967677-53967677 on Build GRCh38
rs776609888 Chr.1: 53967799-53967799 on Build GRCh38
rs747712863 Chr.1: 53967757-53967758 on Build GRCh38
rs148123779 Chr.1: 53967746-53967746 on Build GRCh38
rs1023546324 Chr.1: 53967756-53967756 on Build GRCh38
rs757870981 Chr.1: 53967745-53967745 on Build GRCh38
rs35123335 Chr.1: 53967680-53967680 on Build GRCh38
rs538865173 Chr.1: 53967611-53967611 on Build GRCh38
rs911017111 Chr.1: 53967760-53967761 on Build GRCh38
rs373099914 Chr.1: 53967782-53967782 on Build GRCh38
rs753033638 Chr.1: 53967725-53967725 on Build GRCh38
rs967986267 Chr.1: 53967719-53967719 on Build GRCh38
rs202087838 Chr.1: 53967721-53967721 on Build GRCh38
rs79416338 Chr.1: 53967646-53967646 on Build GRCh38
rs145151431 Chr.1: 53967792-53967792 on Build GRCh38
rs770252097 Chr.1: 53967751-53967751 on Build GRCh38
rs140495032 Chr.1: 53967613-53967613 on Build GRCh38
rs752065781 Chr.1: 53967733-53967733 on Build GRCh38
rs139281324 Chr.1: 53967793-53967793 on Build GRCh38
rs1011352013 Chr.1: 53967748-53967748 on Build GRCh38
rs899694727 Chr.1: 53967612-53967612 on Build GRCh38
rs941179837 Chr.1: 53967589-53967589 on Build GRCh38
rs761214992 Chr.1: 53967679-53967679 on Build GRCh38
rs768160886 Chr.1: 53967674-53967674 on Build GRCh38
rs756193247 Chr.1: 53967765-53967765 on Build GRCh38
rs766111607 Chr.1: 53967712-53967712 on Build GRCh38
rs753661680 Chr.1: 53967720-53967720 on Build GRCh38
rs940924680 Chr.1: 53967611-53967611 on Build GRCh38
rs758343591 Chr.1: 53967726-53967726 on Build GRCh38
rs1031041106 Chr.1: 53967773-53967773 on Build GRCh38
rs1038593664 Chr.1: 53967595-53967595 on Build GRCh38
rs773063752 Chr.1: 53967689-53967689 on Build GRCh38
rs577707844 Chr.1: 53967631-53967631 on Build GRCh38
rs149677498 Chr.1: 53967673-53967673 on Build GRCh38
rs747523700 Chr.1: 53967665-53967665 on Build GRCh38
rs760615054 Chr.1: 53967691-53967691 on Build GRCh38
rs551296330 Chr.1: 53967732-53967732 on Build GRCh38
rs778424436 Chr.1: 53967759-53967759 on Build GRCh38
rs772456393 Chr.1: 53967599-53967599 on Build GRCh38
rs143699160 Chr.1: 53967722-53967722 on Build GRCh38
rs187681558 Chr.1: 53967645-53967645 on Build GRCh38

View Full Product Details