Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 76623070-76623323 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
F2RL2
Entrez Gene ID
2151
Gene Name
coagulation factor II thrombin receptor like 2
Gene Aliases
PAR-3, PAR3
UniGene
Hs.42502
Gene Chromosome Location
Chr.5: 76615482-76623434 on Build GRCh38
Transcripts
NM_001256566.1 NM_004101.3 NM_001285460.1 NM_001285461.1 NM_001285462.1 NM_006633.3 XM_005248410.2 XM_005248414.1 XM_011543108.1 XM_017008960.1

Gene Symbol
IQGAP2
Entrez Gene ID
10788
Gene Name
IQ motif containing GTPase activating protein 2
Gene Aliases
-
UniGene
Hs.291030
Gene Chromosome Location
Chr.5: 76403255-76708132 on Build GRCh38
Transcripts
NM_001256566.1 NM_004101.3 NM_001285460.1 NM_001285461.1 NM_001285462.1 NM_006633.3 XM_005248410.2 XM_005248414.1 XM_011543108.1 XM_017008960.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1722093 c.64+1G>A p.? Chr.5: 76623166-76623166 on Build GRCh38
4951638 c.2T>G p.M1R Chr.5: 76623229-76623229 on Build GRCh38
362339 c.27T>C p.A9A Chr.5: 76623204-76623204 on Build GRCh38
6657725 c.64+2T>C p.? Chr.5: 76623165-76623165 on Build GRCh38
5673232 c.35T>A p.L12Q Chr.5: 76623196-76623196 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs753113982 Chr.5: 76623149-76623149 on Build GRCh38
rs2069649 Chr.5: 76623187-76623187 on Build GRCh38
rs904459722 Chr.5: 76623286-76623286 on Build GRCh38
rs1034162332 Chr.5: 76623290-76623290 on Build GRCh38
rs768780535 Chr.5: 76623248-76623248 on Build GRCh38
rs999948253 Chr.5: 76623289-76623289 on Build GRCh38
rs748377011 Chr.5: 76623183-76623183 on Build GRCh38
rs778389017 Chr.5: 76623212-76623212 on Build GRCh38
rs757378637 Chr.5: 76623183-76623183 on Build GRCh38
rs368602737 Chr.5: 76623300-76623300 on Build GRCh38
rs79814375 Chr.5: 76623148-76623148 on Build GRCh38
rs941502356 Chr.5: 76623151-76623151 on Build GRCh38
rs550954885 Chr.5: 76623267-76623267 on Build GRCh38
rs778793111 Chr.5: 76623249-76623249 on Build GRCh38
rs200798548 Chr.5: 76623260-76623260 on Build GRCh38
rs372661497 Chr.5: 76623259-76623259 on Build GRCh38
rs370122521 Chr.5: 76623131-76623131 on Build GRCh38
rs755921653 Chr.5: 76623145-76623146 on Build GRCh38
rs376660059 Chr.5: 76623126-76623126 on Build GRCh38
rs374397419 Chr.5: 76623106-76623106 on Build GRCh38
rs568472748 Chr.5: 76623185-76623185 on Build GRCh38
rs758488879 Chr.5: 76623157-76623157 on Build GRCh38
rs754540183 Chr.5: 76623200-76623200 on Build GRCh38
rs754323126 Chr.5: 76623121-76623121 on Build GRCh38
rs777637035 Chr.5: 76623193-76623203 on Build GRCh38
rs751550382 Chr.5: 76623170-76623170 on Build GRCh38
rs749206178 Chr.5: 76623229-76623229 on Build GRCh38
rs776636414 Chr.5: 76623109-76623109 on Build GRCh38
rs748970412 Chr.5: 76623202-76623203 on Build GRCh38
rs958416456 Chr.5: 76623292-76623292 on Build GRCh38
rs573758676 Chr.5: 76623168-76623168 on Build GRCh38

View Full Product Details