Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.X: 71537870-71538115 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
OGT
Entrez Gene ID
8473
Gene Name
O-linked N-acetylglucosamine (GlcNAc) transferase
Gene Aliases
HINCUT-1, HRNT1, O-GLCNAC
UniGene
Hs.405410
Gene Chromosome Location
Chr.X: 71533062-71575897 on Build GRCh38
Transcripts
NM_181672.2 NM_181673.2 XM_017029907.1 XM_017029908.1

SNP Detail

SNP ID Chromosome Location
rs769431858 Chr.X: 71537979-71537979 on Build GRCh38
rs750443814 Chr.X: 71538069-71538069 on Build GRCh38
rs761726947 Chr.X: 71538025-71538025 on Build GRCh38
rs867971973 Chr.X: 71538061-71538061 on Build GRCh38
rs377762466 Chr.X: 71537948-71537948 on Build GRCh38
rs373260917 Chr.X: 71538088-71538088 on Build GRCh38
rs760907200 Chr.X: 71538080-71538080 on Build GRCh38
rs762773165 Chr.X: 71538000-71538000 on Build GRCh38
rs775173991 Chr.X: 71537994-71537994 on Build GRCh38
rs768305827 Chr.X: 71538003-71538003 on Build GRCh38
rs948915983 Chr.X: 71538037-71538037 on Build GRCh38
rs35949079 Chr.X: 71537914-71537915 on Build GRCh38
rs745523420 Chr.X: 71537968-71537968 on Build GRCh38
rs149977750 Chr.X: 71538068-71538068 on Build GRCh38
rs368768611 Chr.X: 71538049-71538049 on Build GRCh38
rs139217701 Chr.X: 71537923-71537923 on Build GRCh38
rs770618871 Chr.X: 71537936-71537936 on Build GRCh38
rs774321086 Chr.X: 71538017-71538017 on Build GRCh38
rs200669552 Chr.X: 71538048-71538048 on Build GRCh38

View Full Product Details