Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 9162816-9163063 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
NTN1
Entrez Gene ID
9423
Gene Name
netrin 1
Gene Aliases
NTN1L
UniGene
Hs.660885
Gene Chromosome Location
Chr.17: 9021542-9244000 on Build GRCh38
Transcripts
NM_004822.2 XM_006721595.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6368970 c.1049G>A p.C350Y Chr.17: 9162843-9162843 on Build GRCh38
4878504 c.1129G>A p.G377S Chr.17: 9162923-9162923 on Build GRCh38
5882972 c.1103_1104GC>AA p.C368* Chr.17: 9162897-9162898 on Build GRCh38
7241175 c.1143T>C p.H381H Chr.17: 9162937-9162937 on Build GRCh38
4388276 c.1080G>A p.S360S Chr.17: 9162874-9162874 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs748575791 Chr.17: 9163036-9163036 on Build GRCh38
rs766047025 Chr.17: 9163008-9163008 on Build GRCh38
rs531668666 Chr.17: 9162846-9162846 on Build GRCh38
rs531000196 Chr.17: 9163026-9163026 on Build GRCh38
rs150043655 Chr.17: 9162991-9162991 on Build GRCh38
rs893720110 Chr.17: 9163030-9163030 on Build GRCh38
rs747583915 Chr.17: 9162962-9162962 on Build GRCh38
rs769366933 Chr.17: 9162985-9162985 on Build GRCh38
rs772961270 Chr.17: 9162987-9162987 on Build GRCh38
rs140248343 Chr.17: 9162845-9162845 on Build GRCh38
rs375204917 Chr.17: 9162836-9162836 on Build GRCh38
rs763773917 Chr.17: 9163019-9163019 on Build GRCh38
rs772623927 Chr.17: 9162908-9162908 on Build GRCh38
rs773929461 Chr.17: 9163011-9163011 on Build GRCh38
rs758282081 Chr.17: 9162860-9162860 on Build GRCh38
rs772486953 Chr.17: 9162886-9162886 on Build GRCh38
rs558564980 Chr.17: 9162879-9162879 on Build GRCh38
rs765779295 Chr.17: 9162917-9162917 on Build GRCh38
rs749586755 Chr.17: 9162878-9162878 on Build GRCh38
rs761444091 Chr.17: 9162841-9162841 on Build GRCh38
rs921898990 Chr.17: 9163035-9163035 on Build GRCh38
rs370864994 Chr.17: 9162835-9162835 on Build GRCh38
rs779997082 Chr.17: 9162863-9162863 on Build GRCh38
rs375467976 Chr.17: 9162874-9162874 on Build GRCh38
rs769085897 Chr.17: 9162977-9162977 on Build GRCh38
rs1037160564 Chr.17: 9162936-9162936 on Build GRCh38
rs758553071 Chr.17: 9162960-9162960 on Build GRCh38
rs751598491 Chr.17: 9162919-9162919 on Build GRCh38
rs776977895 Chr.17: 9162903-9162903 on Build GRCh38
rs755497852 Chr.17: 9163028-9163028 on Build GRCh38
rs776994544 Chr.17: 9162979-9162979 on Build GRCh38
rs762489133 Chr.17: 9162915-9162915 on Build GRCh38
rs757439358 Chr.17: 9162926-9162926 on Build GRCh38
rs199843845 Chr.17: 9163022-9163022 on Build GRCh38
rs112161714 Chr.17: 9162922-9162922 on Build GRCh38
rs754959235 Chr.17: 9162920-9162920 on Build GRCh38
rs748971464 Chr.17: 9163043-9163043 on Build GRCh38
rs781637615 Chr.17: 9163032-9163032 on Build GRCh38
rs144950762 Chr.17: 9162904-9162904 on Build GRCh38
rs115807192 Chr.17: 9163040-9163040 on Build GRCh38
rs200276891 Chr.17: 9163017-9163017 on Build GRCh38
rs368406936 Chr.17: 9162961-9162961 on Build GRCh38
rs374707235 Chr.17: 9162840-9162840 on Build GRCh38
rs751434453 Chr.17: 9162864-9162864 on Build GRCh38
rs767509435 Chr.17: 9163015-9163015 on Build GRCh38
rs754863907 Chr.17: 9162869-9162869 on Build GRCh38
rs767421459 Chr.17: 9162921-9162921 on Build GRCh38
rs779443102 Chr.17: 9163023-9163024 on Build GRCh38
rs932952042 Chr.17: 9163009-9163009 on Build GRCh38
rs778868022 Chr.17: 9162883-9162883 on Build GRCh38
rs368127099 Chr.17: 9162837-9162837 on Build GRCh38
rs755976695 Chr.17: 9163018-9163018 on Build GRCh38
rs748496958 Chr.17: 9162982-9162982 on Build GRCh38
rs55687153 Chr.17: 9162931-9162931 on Build GRCh38
rs8067722 Chr.17: 9163012-9163012 on Build GRCh38
rs2286511 Chr.17: 9162937-9162937 on Build GRCh38
rs181255712 Chr.17: 9163044-9163044 on Build GRCh38
rs563402359 Chr.17: 9163023-9163023 on Build GRCh38
rs769069239 Chr.17: 9162898-9162898 on Build GRCh38

View Full Product Details