Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 139363718-139363979 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PAIP2
Entrez Gene ID
51247
Gene Name
poly(A) binding protein interacting protein 2
Gene Aliases
PAIP-2, PAIP2A
UniGene
Hs.396644
Gene Chromosome Location
Chr.5: 139341832-139369720 on Build GRCh38
Transcripts
NM_001033112.2 NM_016480.4 NR_109830.1 XM_011543428.1 XM_017009522.1 XM_017009523.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6908140 c.82G>C p.D28H Chr.5: 139363866-139363866 on Build GRCh38
287861 c.84C>G p.D28E Chr.5: 139363868-139363868 on Build GRCh38
6036991 c.86A>G p.N29S Chr.5: 139363870-139363870 on Build GRCh38
448670 c.63C>T p.N21N Chr.5: 139363847-139363847 on Build GRCh38
6284102 c.130A>C p.N44H Chr.5: 139363914-139363914 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs972143090 Chr.5: 139363952-139363952 on Build GRCh38
rs371548085 Chr.5: 139363849-139363849 on Build GRCh38
rs966511538 Chr.5: 139363928-139363928 on Build GRCh38
rs11548835 Chr.5: 139363914-139363914 on Build GRCh38
rs377311638 Chr.5: 139363889-139363889 on Build GRCh38
rs753242971 Chr.5: 139363882-139363882 on Build GRCh38
rs775318653 Chr.5: 139363791-139363791 on Build GRCh38
rs927491251 Chr.5: 139363805-139363805 on Build GRCh38
rs11548840 Chr.5: 139363815-139363815 on Build GRCh38
rs751922665 Chr.5: 139363835-139363835 on Build GRCh38
rs372634342 Chr.5: 139363943-139363943 on Build GRCh38
rs945712388 Chr.5: 139363852-139363852 on Build GRCh38
rs34490218 Chr.5: 139363816-139363817 on Build GRCh38
rs763200663 Chr.5: 139363829-139363829 on Build GRCh38
rs201557008 Chr.5: 139363879-139363879 on Build GRCh38
rs576725648 Chr.5: 139363800-139363800 on Build GRCh38
rs746831677 Chr.5: 139363792-139363792 on Build GRCh38
rs755406749 Chr.5: 139363847-139363847 on Build GRCh38
rs748749763 Chr.5: 139363768-139363768 on Build GRCh38
rs375110036 Chr.5: 139363773-139363773 on Build GRCh38
rs1009259842 Chr.5: 139363759-139363759 on Build GRCh38
rs1057506035 Chr.5: 139363821-139363821 on Build GRCh38
rs778479608 Chr.5: 139363858-139363858 on Build GRCh38
rs368501300 Chr.5: 139363940-139363940 on Build GRCh38
rs201030798 Chr.5: 139363880-139363880 on Build GRCh38
rs199645765 Chr.5: 139363848-139363848 on Build GRCh38
rs552998100 Chr.5: 139363942-139363942 on Build GRCh38
rs776558495 Chr.5: 139363801-139363801 on Build GRCh38
rs768069937 Chr.5: 139363859-139363859 on Build GRCh38
rs199918986 Chr.5: 139363834-139363834 on Build GRCh38
rs745398548 Chr.5: 139363907-139363907 on Build GRCh38
rs765343045 Chr.5: 139363811-139363811 on Build GRCh38
rs779691552 Chr.5: 139363941-139363941 on Build GRCh38
rs866324937 Chr.5: 139363820-139363820 on Build GRCh38
rs781769945 Chr.5: 139363755-139363755 on Build GRCh38
rs771664296 Chr.5: 139363780-139363780 on Build GRCh38
rs773211784 Chr.5: 139363828-139363828 on Build GRCh38
rs745413402 Chr.5: 139363942-139363943 on Build GRCh38
rs756627525 Chr.5: 139363886-139363886 on Build GRCh38
rs141171789 Chr.5: 139363810-139363810 on Build GRCh38
rs367802509 Chr.5: 139363776-139363776 on Build GRCh38
rs113267132 Chr.5: 139363770-139363770 on Build GRCh38
rs1051614684 Chr.5: 139363841-139363841 on Build GRCh38

View Full Product Details