Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 19338752-19339023 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
MAU2
Entrez Gene ID
23383
Gene Name
MAU2 sister chromatid cohesion factor
Gene Aliases
KIAA0892, MAU2L, SCC4, mau-2
UniGene
Hs.654939
Gene Chromosome Location
Chr.19: 19320686-19358754 on Build GRCh38
Transcripts
NM_015329.3 XM_005259837.3 XM_005259838.3 XM_006722711.3 XM_011527844.2 XM_017026539.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
382076 c.478G>C p.D160H Chr.19: 19338866-19338866 on Build GRCh38
6708916 c.528G>A p.R176R Chr.19: 19338916-19338916 on Build GRCh38
6250891 c.551+5G>T p.? Chr.19: 19338944-19338944 on Build GRCh38
993322 c.490G>A p.A164T Chr.19: 19338878-19338878 on Build GRCh38
4837939 c.468G>A p.T156T Chr.19: 19338856-19338856 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1057027006 Chr.19: 19338787-19338787 on Build GRCh38
rs765291205 Chr.19: 19338966-19338966 on Build GRCh38
rs763952936 Chr.19: 19338872-19338872 on Build GRCh38
rs766518453 Chr.19: 19338848-19338848 on Build GRCh38
rs200212304 Chr.19: 19338907-19338907 on Build GRCh38
rs117955413 Chr.19: 19338910-19338910 on Build GRCh38
rs542197684 Chr.19: 19338871-19338871 on Build GRCh38
rs915182763 Chr.19: 19338969-19338969 on Build GRCh38
rs760801458 Chr.19: 19338838-19338838 on Build GRCh38
rs377473235 Chr.19: 19338886-19338886 on Build GRCh38
rs375537806 Chr.19: 19338916-19338916 on Build GRCh38
rs946662589 Chr.19: 19338972-19338972 on Build GRCh38
rs776697509 Chr.19: 19338952-19338952 on Build GRCh38
rs773540131 Chr.19: 19338818-19338818 on Build GRCh38
rs767397209 Chr.19: 19338982-19338982 on Build GRCh38
rs772553325 Chr.19: 19338938-19338938 on Build GRCh38
rs751567233 Chr.19: 19338978-19338978 on Build GRCh38
rs988029302 Chr.19: 19338964-19338964 on Build GRCh38
rs2074090 Chr.19: 19338877-19338877 on Build GRCh38
rs757105871 Chr.19: 19338874-19338874 on Build GRCh38
rs748764235 Chr.19: 19338913-19338913 on Build GRCh38
rs759554169 Chr.19: 19338955-19338955 on Build GRCh38
rs760981254 Chr.19: 19338948-19338948 on Build GRCh38
rs750134520 Chr.19: 19338983-19338983 on Build GRCh38
rs774941172 Chr.19: 19338922-19338922 on Build GRCh38
rs368252240 Chr.19: 19338934-19338934 on Build GRCh38
rs956596960 Chr.19: 19338963-19338963 on Build GRCh38
rs762947033 Chr.19: 19338904-19338904 on Build GRCh38
rs928325879 Chr.19: 19338786-19338786 on Build GRCh38
rs1028663496 Chr.19: 19338895-19338895 on Build GRCh38
rs773416738 Chr.19: 19338947-19338947 on Build GRCh38
rs981381087 Chr.19: 19338781-19338781 on Build GRCh38
rs761627733 Chr.19: 19338979-19338979 on Build GRCh38
rs199781947 Chr.19: 19338856-19338856 on Build GRCh38
rs2074089 Chr.19: 19338794-19338794 on Build GRCh38

View Full Product Details