Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 172686048-172686321 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
NEURL1B
Entrez Gene ID
54492
Gene Name
neuralized E3 ubiquitin protein ligase 1B
Gene Aliases
NEURL3, hNeur2, neur2
UniGene
Hs.91521
Gene Chromosome Location
Chr.5: 172641266-172691540 on Build GRCh38
Transcripts
NM_001142651.2 NM_001308177.1 NM_001308178.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5597162 c.1341C>T p.S447S Chr.5: 172686214-172686214 on Build GRCh38
3761110 c.1359C>T p.D453D Chr.5: 172686232-172686232 on Build GRCh38
1065863 c.1314C>T p.S438S Chr.5: 172686187-172686187 on Build GRCh38
3744780 c.1423+8A>T p.? Chr.5: 172686304-172686304 on Build GRCh38
1065865 c.1325C>T p.T442M Chr.5: 172686198-172686198 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs886712069 Chr.5: 172686070-172686070 on Build GRCh38
rs779539166 Chr.5: 172686269-172686271 on Build GRCh38
rs1047992773 Chr.5: 172686069-172686069 on Build GRCh38
rs768527288 Chr.5: 172686248-172686248 on Build GRCh38
rs1002919399 Chr.5: 172686198-172686198 on Build GRCh38
rs894085813 Chr.5: 172686111-172686111 on Build GRCh38
rs1053956051 Chr.5: 172686077-172686077 on Build GRCh38
rs549973149 Chr.5: 172686194-172686194 on Build GRCh38
rs1016850282 Chr.5: 172686107-172686107 on Build GRCh38
rs778473251 Chr.5: 172686192-172686192 on Build GRCh38
rs756883063 Chr.5: 172686138-172686138 on Build GRCh38
rs552398934 Chr.5: 172686276-172686276 on Build GRCh38
rs566022020 Chr.5: 172686277-172686277 on Build GRCh38
rs532298637 Chr.5: 172686233-172686233 on Build GRCh38
rs913796259 Chr.5: 172686204-172686204 on Build GRCh38
rs979250287 Chr.5: 172686269-172686269 on Build GRCh38
rs747829 Chr.5: 172686232-172686232 on Build GRCh38
rs369713049 Chr.5: 172686214-172686214 on Build GRCh38
rs373249276 Chr.5: 172686221-172686221 on Build GRCh38
rs17075071 Chr.5: 172686071-172686071 on Build GRCh38
rs1009807675 Chr.5: 172686132-172686132 on Build GRCh38
rs996701998 Chr.5: 172686118-172686118 on Build GRCh38
rs534902647 Chr.5: 172686293-172686293 on Build GRCh38
rs1021595167 Chr.5: 172686139-172686139 on Build GRCh38

View Full Product Details