Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 42721862-42722103 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PRPH2
Entrez Gene ID
5961
Gene Name
peripherin 2
Gene Aliases
AOFMD, AVMD, CACD2, DS, MDBS1, PRPH, RDS, RP7, TSPAN22, rd2
UniGene
Hs.654489
Gene Chromosome Location
Chr.6: 42694509-42723024 on Build GRCh38
Transcripts
NM_000322.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4718043 c.455T>C p.M152T Chr.6: 42721880-42721880 on Build GRCh38
3394204 c.425G>A p.R142Q Chr.6: 42721910-42721910 on Build GRCh38
4718044 c.244T>A p.C82S Chr.6: 42722091-42722091 on Build GRCh38
188502 c.469G>A p.D157N Chr.6: 42721866-42721866 on Build GRCh38
6218978 c.408C>T p.N136N Chr.6: 42721927-42721927 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs750164572 Chr.6: 42721890-42721890 on Build GRCh38
rs121918561 Chr.6: 42721981-42721983 on Build GRCh38
rs150695654 Chr.6: 42722035-42722035 on Build GRCh38
rs148513859 Chr.6: 42721927-42721927 on Build GRCh38
rs672601326 Chr.6: 42721913-42721914 on Build GRCh38
rs200978519 Chr.6: 42721900-42721900 on Build GRCh38
rs781631103 Chr.6: 42722014-42722014 on Build GRCh38
rs772861671 Chr.6: 42721961-42721961 on Build GRCh38
rs530135649 Chr.6: 42722065-42722065 on Build GRCh38
rs771068866 Chr.6: 42721923-42721923 on Build GRCh38
rs768473101 Chr.6: 42721938-42721938 on Build GRCh38
rs7764439 Chr.6: 42722017-42722017 on Build GRCh38
rs554945964 Chr.6: 42721910-42721910 on Build GRCh38
rs375823961 Chr.6: 42721897-42721897 on Build GRCh38
rs61755779 Chr.6: 42721959-42721959 on Build GRCh38
rs772157862 Chr.6: 42722066-42722066 on Build GRCh38
rs770029839 Chr.6: 42721930-42721930 on Build GRCh38
rs950303632 Chr.6: 42722011-42722011 on Build GRCh38
rs574930557 Chr.6: 42721933-42721933 on Build GRCh38
rs774210091 Chr.6: 42721954-42721954 on Build GRCh38
rs543703718 Chr.6: 42721955-42721955 on Build GRCh38
rs773254206 Chr.6: 42722070-42722070 on Build GRCh38
rs768400169 Chr.6: 42722036-42722036 on Build GRCh38
rs760311433 Chr.6: 42722082-42722082 on Build GRCh38
rs61755782 Chr.6: 42721911-42721912 on Build GRCh38
rs760213205 Chr.6: 42721966-42721966 on Build GRCh38
rs140227298 Chr.6: 42721989-42721989 on Build GRCh38
rs61755781 Chr.6: 42721913-42721913 on Build GRCh38
rs61755776 Chr.6: 42722032-42722032 on Build GRCh38
rs753035269 Chr.6: 42721898-42721898 on Build GRCh38
rs563581127 Chr.6: 42721968-42721968 on Build GRCh38
rs139936445 Chr.6: 42722083-42722083 on Build GRCh38
rs532319562 Chr.6: 42721986-42721986 on Build GRCh38
rs527236097 Chr.6: 42721925-42721925 on Build GRCh38
rs769723975 Chr.6: 42721941-42721941 on Build GRCh38
rs747893076 Chr.6: 42722064-42722064 on Build GRCh38
rs61755780 Chr.6: 42721914-42721914 on Build GRCh38
rs61755784 Chr.6: 42721894-42721894 on Build GRCh38
rs567678322 Chr.6: 42722055-42722055 on Build GRCh38
rs61755783 Chr.6: 42721911-42721911 on Build GRCh38
rs200009675 Chr.6: 42722023-42722023 on Build GRCh38
rs753507738 Chr.6: 42721972-42721972 on Build GRCh38
rs777764923 Chr.6: 42721960-42721960 on Build GRCh38
rs762692183 Chr.6: 42721936-42721936 on Build GRCh38
rs61755778 Chr.6: 42721964-42721964 on Build GRCh38
rs781256236 Chr.6: 42721926-42721926 on Build GRCh38
rs61755777 Chr.6: 42721978-42721980 on Build GRCh38
rs757778182 Chr.6: 42722001-42722001 on Build GRCh38

View Full Product Details