Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.X: 103825027-103825288 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
RAB9B
Entrez Gene ID
51209
Gene Name
RAB9B, member RAS oncogene family
Gene Aliases
RAB9L, Rab-9L
UniGene
Hs.522736
Gene Chromosome Location
Chr.X: 103822325-103832282 on Build GRCh38
Transcripts
NM_016370.2

SNP Detail

SNP ID Chromosome Location
rs200974641 Chr.X: 103825152-103825152 on Build GRCh38
rs536203957 Chr.X: 103825097-103825097 on Build GRCh38
rs761856500 Chr.X: 103825189-103825189 on Build GRCh38
rs368010774 Chr.X: 103825213-103825213 on Build GRCh38
rs749040974 Chr.X: 103825165-103825165 on Build GRCh38
rs375103308 Chr.X: 103825140-103825140 on Build GRCh38
rs768904031 Chr.X: 103825178-103825178 on Build GRCh38
rs199638729 Chr.X: 103825269-103825269 on Build GRCh38
rs745819137 Chr.X: 103825139-103825139 on Build GRCh38
rs759951506 Chr.X: 103825260-103825260 on Build GRCh38
rs763892219 Chr.X: 103825263-103825263 on Build GRCh38
rs747517766 Chr.X: 103825093-103825093 on Build GRCh38
rs754287809 Chr.X: 103825257-103825257 on Build GRCh38
rs142096547 Chr.X: 103825188-103825188 on Build GRCh38
rs1044793623 Chr.X: 103825194-103825194 on Build GRCh38
rs772188064 Chr.X: 103825202-103825202 on Build GRCh38
rs779844580 Chr.X: 103825158-103825158 on Build GRCh38
rs761814878 Chr.X: 103825242-103825242 on Build GRCh38
rs766945249 Chr.X: 103825248-103825248 on Build GRCh38
rs2232731 Chr.X: 103825092-103825092 on Build GRCh38

View Full Product Details