Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.20: 63643999-63644270 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
STMN3
Entrez Gene ID
50861
Gene Name
stathmin 3
Gene Aliases
SCLIP
UniGene
Hs.639609
Gene Chromosome Location
Chr.20: 63639705-63653610 on Build GRCh38
Transcripts
NM_001276310.1 NM_015894.3 NR_075070.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6422233 c.115G>T p.D39Y Chr.20: 63644214-63644214 on Build GRCh38
5974165 c.91C>G p.P31A Chr.20: 63644238-63644238 on Build GRCh38
3363374 c.93C>T p.P31P Chr.20: 63644236-63644236 on Build GRCh38
5365918 c.87G>A p.P29P Chr.20: 63644242-63644242 on Build GRCh38
4632843 c.73T>C p.F25L Chr.20: 63644256-63644256 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs754452436 Chr.20: 63644171-63644196 on Build GRCh38
rs753296813 Chr.20: 63644167-63644168 on Build GRCh38
rs994205499 Chr.20: 63644070-63644070 on Build GRCh38
rs751010777 Chr.20: 63644036-63644036 on Build GRCh38
rs758782392 Chr.20: 63644050-63644050 on Build GRCh38
rs762539489 Chr.20: 63644205-63644205 on Build GRCh38
rs897043368 Chr.20: 63644170-63644170 on Build GRCh38
rs770151991 Chr.20: 63644243-63644243 on Build GRCh38
rs570633127 Chr.20: 63644125-63644125 on Build GRCh38
rs372313701 Chr.20: 63644196-63644196 on Build GRCh38
rs746092434 Chr.20: 63644181-63644181 on Build GRCh38
rs751683926 Chr.20: 63644238-63644238 on Build GRCh38
rs759108217 Chr.20: 63644217-63644217 on Build GRCh38
rs750619499 Chr.20: 63644230-63644230 on Build GRCh38
rs755267296 Chr.20: 63644197-63644197 on Build GRCh38
rs762464629 Chr.20: 63644163-63644163 on Build GRCh38
rs372125936 Chr.20: 63644192-63644192 on Build GRCh38
rs760406271 Chr.20: 63644227-63644227 on Build GRCh38
rs758526098 Chr.20: 63644236-63644236 on Build GRCh38
rs765826343 Chr.20: 63644164-63644164 on Build GRCh38
rs765186968 Chr.20: 63644162-63644162 on Build GRCh38
rs376628586 Chr.20: 63644167-63644167 on Build GRCh38
rs888537333 Chr.20: 63644130-63644130 on Build GRCh38
rs766099123 Chr.20: 63644215-63644215 on Build GRCh38
rs557013511 Chr.20: 63644051-63644051 on Build GRCh38
rs764857512 Chr.20: 63644172-63644173 on Build GRCh38
rs754883366 Chr.20: 63644172-63644172 on Build GRCh38
rs374011984 Chr.20: 63644180-63644180 on Build GRCh38
rs568352689 Chr.20: 63644027-63644027 on Build GRCh38
rs748684575 Chr.20: 63644242-63644242 on Build GRCh38
rs758456743 Chr.20: 63644183-63644183 on Build GRCh38
rs926114173 Chr.20: 63644086-63644086 on Build GRCh38
rs111797439 Chr.20: 63644075-63644075 on Build GRCh38
rs141932828 Chr.20: 63644176-63644176 on Build GRCh38
rs202108425 Chr.20: 63644240-63644240 on Build GRCh38
rs767441107 Chr.20: 63644218-63644218 on Build GRCh38
rs755972157 Chr.20: 63644177-63644177 on Build GRCh38
rs947098772 Chr.20: 63644031-63644031 on Build GRCh38
rs970317669 Chr.20: 63644146-63644146 on Build GRCh38
rs537081157 Chr.20: 63644030-63644030 on Build GRCh38
rs772868182 Chr.20: 63644202-63644202 on Build GRCh38
rs780380420 Chr.20: 63644237-63644237 on Build GRCh38
rs6010985 Chr.20: 63644139-63644139 on Build GRCh38
rs889558197 Chr.20: 63644160-63644160 on Build GRCh38
rs756907593 Chr.20: 63644062-63644062 on Build GRCh38
rs748453793 Chr.20: 63644198-63644198 on Build GRCh38
rs556068883 Chr.20: 63644229-63644229 on Build GRCh38
rs200355112 Chr.20: 63644201-63644201 on Build GRCh38
rs1049932566 Chr.20: 63644032-63644032 on Build GRCh38
rs773832800 Chr.20: 63644216-63644216 on Build GRCh38
rs536342957 Chr.20: 63644133-63644133 on Build GRCh38

View Full Product Details