Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 6481915-6482140 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
UHRF2
Entrez Gene ID
115426
Gene Name
ubiquitin like with PHD and ring finger domains 2
Gene Aliases
NIRF, RNF107, TDRD23, URF2
UniGene
Hs.493401
Gene Chromosome Location
Chr.9: 6413148-6508108 on Build GRCh38
Transcripts
NM_152896.2 NR_046386.1 XM_011517704.2 XM_011517705.2 XM_017014253.1 XM_017014254.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
32661 c.1392+3A>G p.? Chr.9: 6482102-6482102 on Build GRCh38
4739146 c.1307C>T p.T436M Chr.9: 6482014-6482014 on Build GRCh38
5698597 c.1377G>T p.W459C Chr.9: 6482084-6482084 on Build GRCh38
6423376 c.1376G>T p.W459L Chr.9: 6482083-6482083 on Build GRCh38
1109531 c.1389T>C p.V463V Chr.9: 6482096-6482096 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs766214915 Chr.9: 6482057-6482057 on Build GRCh38
rs756763305 Chr.9: 6481949-6481949 on Build GRCh38
rs756144819 Chr.9: 6482044-6482044 on Build GRCh38
rs566056050 Chr.9: 6481970-6481970 on Build GRCh38
rs753669590 Chr.9: 6482069-6482069 on Build GRCh38
rs79300900 Chr.9: 6482002-6482002 on Build GRCh38
rs372282086 Chr.9: 6482087-6482087 on Build GRCh38
rs529815356 Chr.9: 6481972-6481972 on Build GRCh38
rs888055127 Chr.9: 6482111-6482111 on Build GRCh38
rs767186246 Chr.9: 6482019-6482019 on Build GRCh38
rs781440660 Chr.9: 6482117-6482118 on Build GRCh38
rs749033730 Chr.9: 6481965-6481965 on Build GRCh38
rs761434516 Chr.9: 6482015-6482015 on Build GRCh38
rs372041814 Chr.9: 6481941-6481941 on Build GRCh38
rs763167150 Chr.9: 6482010-6482010 on Build GRCh38
rs377446685 Chr.9: 6481953-6481953 on Build GRCh38
rs781313503 Chr.9: 6481950-6481950 on Build GRCh38
rs763845489 Chr.9: 6482011-6482011 on Build GRCh38
rs749869662 Chr.9: 6482027-6482027 on Build GRCh38
rs747121284 Chr.9: 6481982-6481982 on Build GRCh38
rs375783512 Chr.9: 6482116-6482116 on Build GRCh38
rs150054333 Chr.9: 6482079-6482079 on Build GRCh38
rs748757857 Chr.9: 6481985-6481985 on Build GRCh38
rs368717130 Chr.9: 6481983-6481983 on Build GRCh38
rs750340298 Chr.9: 6481951-6481951 on Build GRCh38
rs61760197 Chr.9: 6482100-6482100 on Build GRCh38
rs751221575 Chr.9: 6482014-6482014 on Build GRCh38
rs568891600 Chr.9: 6481959-6481959 on Build GRCh38
rs772396676 Chr.9: 6481971-6481971 on Build GRCh38
rs755983315 Chr.9: 6481955-6481955 on Build GRCh38

View Full Product Details