Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 129136460-129136723 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PTPA
Entrez Gene ID
5524
Gene Name
protein phosphatase 2 phosphatase activator
Gene Aliases
PP2A, PPP2R4, PR53
UniGene
Hs.400740
Gene Chromosome Location
Chr.9: 129110949-129148946 on Build GRCh38
Transcripts
NM_001193397.1 NM_001271832.1 NM_021131.4 NM_178000.2 NM_178001.2 NM_178003.2 XM_011518834.2 XM_011518835.2 XM_011518836.2 XM_011518837.2 XM_011518838.2 XM_017014888.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6550512 c.673G>T p.E225* Chr.9: 129136478-129136478 on Build GRCh38
3847819 c.760C>T p.P254S Chr.9: 129136565-129136565 on Build GRCh38
397909 c.718G>A p.G240S Chr.9: 129136523-129136523 on Build GRCh38
6754380 c.668A>G p.Y223C Chr.9: 129136473-129136473 on Build GRCh38
1624608 c.682C>T p.R228W Chr.9: 129136487-129136487 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs112063689 Chr.9: 129136595-129136595 on Build GRCh38
rs973370211 Chr.9: 129136617-129136617 on Build GRCh38
rs541420565 Chr.9: 129136629-129136629 on Build GRCh38
rs1003357198 Chr.9: 129136675-129136675 on Build GRCh38
rs775455906 Chr.9: 129136664-129136664 on Build GRCh38
rs201651829 Chr.9: 129136545-129136545 on Build GRCh38
rs748930542 Chr.9: 129136507-129136507 on Build GRCh38
rs374650004 Chr.9: 129136591-129136591 on Build GRCh38
rs1034463504 Chr.9: 129136684-129136684 on Build GRCh38
rs748905012 Chr.9: 129136639-129136639 on Build GRCh38
rs561673686 Chr.9: 129136656-129136656 on Build GRCh38
rs764692216 Chr.9: 129136663-129136663 on Build GRCh38
rs954227908 Chr.9: 129136523-129136523 on Build GRCh38
rs756426357 Chr.9: 129136574-129136574 on Build GRCh38
rs959444370 Chr.9: 129136685-129136687 on Build GRCh38
rs752890749 Chr.9: 129136605-129136605 on Build GRCh38
rs773117020 Chr.9: 129136585-129136585 on Build GRCh38
rs1056136270 Chr.9: 129136685-129136685 on Build GRCh38
rs528116881 Chr.9: 129136626-129136626 on Build GRCh38
rs17508881 Chr.9: 129136522-129136522 on Build GRCh38
rs141469321 Chr.9: 129136573-129136573 on Build GRCh38
rs1012097419 Chr.9: 129136686-129136686 on Build GRCh38
rs369013775 Chr.9: 129136614-129136614 on Build GRCh38
rs141815672 Chr.9: 129136588-129136588 on Build GRCh38
rs759748940 Chr.9: 129136598-129136598 on Build GRCh38
rs530697759 Chr.9: 129136657-129136657 on Build GRCh38
rs754033690 Chr.9: 129136593-129136593 on Build GRCh38
rs1022189501 Chr.9: 129136689-129136689 on Build GRCh38
rs937707802 Chr.9: 129136679-129136679 on Build GRCh38
rs35481042 Chr.9: 129136498-129136498 on Build GRCh38
rs773852913 Chr.9: 129136549-129136549 on Build GRCh38
rs758397055 Chr.9: 129136606-129136606 on Build GRCh38
rs1536548 Chr.9: 129136601-129136601 on Build GRCh38
rs771401589 Chr.9: 129136493-129136493 on Build GRCh38
rs778424213 Chr.9: 129136643-129136643 on Build GRCh38
rs531763781 Chr.9: 129136661-129136661 on Build GRCh38
rs923647865 Chr.9: 129136495-129136495 on Build GRCh38
rs143681457 Chr.9: 129136489-129136489 on Build GRCh38
rs576896363 Chr.9: 129136567-129136567 on Build GRCh38
rs754590808 Chr.9: 129136642-129136642 on Build GRCh38
rs779894271 Chr.9: 129136504-129136504 on Build GRCh38

View Full Product Details