Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 81070389-81070640 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
RASGRF2
Entrez Gene ID
5924
Gene Name
Ras protein specific guanine nucleotide releasing factor 2
Gene Aliases
GRF2, RAS-GRF2
UniGene
Hs.162129
Gene Chromosome Location
Chr.5: 80960672-81230162 on Build GRCh38
Transcripts
NM_006909.2 XM_005248565.1 XM_017009682.1 XM_017009683.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6470541 c.610C>T p.P204S Chr.5: 81070558-81070558 on Build GRCh38
1223223 c.604G>A p.E202K Chr.5: 81070552-81070552 on Build GRCh38
289549 c.617T>C p.I206T Chr.5: 81070565-81070565 on Build GRCh38
1620604 c.621G>T p.K207N Chr.5: 81070569-81070569 on Build GRCh38
6763257 c.601G>T p.D201Y Chr.5: 81070549-81070549 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs376087567 Chr.5: 81070492-81070492 on Build GRCh38
rs762123538 Chr.5: 81070503-81070503 on Build GRCh38
rs760768979 Chr.5: 81070516-81070516 on Build GRCh38
rs756757856 Chr.5: 81070443-81070443 on Build GRCh38
rs377520116 Chr.5: 81070588-81070588 on Build GRCh38
rs780712139 Chr.5: 81070452-81070452 on Build GRCh38
rs762706337 Chr.5: 81070534-81070534 on Build GRCh38
rs749767090 Chr.5: 81070610-81070610 on Build GRCh38
rs746653275 Chr.5: 81070435-81070435 on Build GRCh38
rs183574979 Chr.5: 81070463-81070463 on Build GRCh38
rs376296182 Chr.5: 81070608-81070608 on Build GRCh38
rs745409987 Chr.5: 81070613-81070613 on Build GRCh38
rs138463426 Chr.5: 81070512-81070512 on Build GRCh38
rs752665731 Chr.5: 81070526-81070526 on Build GRCh38
rs1002771799 Chr.5: 81070549-81070549 on Build GRCh38
rs756926154 Chr.5: 81070552-81070552 on Build GRCh38
rs368800371 Chr.5: 81070551-81070551 on Build GRCh38
rs879121333 Chr.5: 81070473-81070473 on Build GRCh38
rs896223015 Chr.5: 81070586-81070586 on Build GRCh38
rs745336585 Chr.5: 81070456-81070456 on Build GRCh38
rs373573174 Chr.5: 81070595-81070595 on Build GRCh38
rs769039334 Chr.5: 81070474-81070474 on Build GRCh38
rs745612142 Chr.5: 81070431-81070431 on Build GRCh38
rs905384220 Chr.5: 81070542-81070542 on Build GRCh38
rs780768973 Chr.5: 81070560-81070560 on Build GRCh38
rs1056251948 Chr.5: 81070577-81070577 on Build GRCh38
rs200881583 Chr.5: 81070545-81070545 on Build GRCh38
rs61759868 Chr.5: 81070497-81070497 on Build GRCh38
rs749942599 Chr.5: 81070590-81070590 on Build GRCh38
rs370420964 Chr.5: 81070615-81070615 on Build GRCh38
rs376557330 Chr.5: 81070525-81070525 on Build GRCh38
rs1035383882 Chr.5: 81070569-81070569 on Build GRCh38
rs913869927 Chr.5: 81070479-81070479 on Build GRCh38
rs770302570 Chr.5: 81070461-81070461 on Build GRCh38
rs772368426 Chr.5: 81070504-81070504 on Build GRCh38
rs374131125 Chr.5: 81070616-81070616 on Build GRCh38
rs749684744 Chr.5: 81070464-81070464 on Build GRCh38

View Full Product Details