Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 81917641-81917890 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SIRT7
Entrez Gene ID
51547
Gene Name
sirtuin 7
Gene Aliases
SIR2L7
UniGene
Hs.514636
Gene Chromosome Location
Chr.17: 81911939-81918182 on Build GRCh38
Transcripts
NM_016538.2 XM_011523580.1 XM_011523581.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6538860 c.213G>C p.L71L Chr.17: 81917848-81917848 on Build GRCh38
403365 c.192G>T p.R64R Chr.17: 81917869-81917869 on Build GRCh38
6562937 c.283G>A p.A95T Chr.17: 81917668-81917668 on Build GRCh38
5988958 c.259G>T p.G87W Chr.17: 81917692-81917692 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs1037478729 Chr.17: 81917860-81917860 on Build GRCh38
rs200237563 Chr.17: 81917720-81917720 on Build GRCh38
rs199694458 Chr.17: 81917781-81917781 on Build GRCh38
rs914933038 Chr.17: 81917800-81917800 on Build GRCh38
rs761836902 Chr.17: 81917796-81917796 on Build GRCh38
rs946382391 Chr.17: 81917818-81917818 on Build GRCh38
rs780026134 Chr.17: 81917851-81917851 on Build GRCh38
rs939058231 Chr.17: 81917848-81917848 on Build GRCh38
rs780368143 Chr.17: 81917691-81917691 on Build GRCh38
rs904954659 Chr.17: 81917828-81917828 on Build GRCh38
rs202037220 Chr.17: 81917718-81917718 on Build GRCh38
rs941493179 Chr.17: 81917801-81917801 on Build GRCh38
rs966936754 Chr.17: 81917680-81917680 on Build GRCh38
rs990411177 Chr.17: 81917795-81917795 on Build GRCh38
rs756369194 Chr.17: 81917671-81917671 on Build GRCh38
rs927925204 Chr.17: 81917738-81917738 on Build GRCh38
rs750503923 Chr.17: 81917802-81917802 on Build GRCh38
rs955429489 Chr.17: 81917736-81917736 on Build GRCh38
rs577056649 Chr.17: 81917864-81917864 on Build GRCh38
rs767601500 Chr.17: 81917797-81917797 on Build GRCh38
rs868456890 Chr.17: 81917758-81917758 on Build GRCh38
rs986948670 Chr.17: 81917742-81917742 on Build GRCh38
rs899034236 Chr.17: 81917870-81917870 on Build GRCh38
rs769460154 Chr.17: 81917749-81917750 on Build GRCh38
rs934918222 Chr.17: 81917770-81917770 on Build GRCh38
rs939348690 Chr.17: 81917739-81917739 on Build GRCh38
rs759096341 Chr.17: 81917735-81917735 on Build GRCh38
rs774595294 Chr.17: 81917716-81917716 on Build GRCh38
rs754022926 Chr.17: 81917857-81917857 on Build GRCh38
rs566414311 Chr.17: 81917764-81917764 on Build GRCh38
rs558814484 Chr.17: 81917813-81917813 on Build GRCh38
rs1044863080 Chr.17: 81917820-81917820 on Build GRCh38
rs138673054 Chr.17: 81917669-81917669 on Build GRCh38
rs776091169 Chr.17: 81917734-81917734 on Build GRCh38
rs769022601 Chr.17: 81917707-81917707 on Build GRCh38
rs749492299 Chr.17: 81917699-81917699 on Build GRCh38
rs747808495 Chr.17: 81917686-81917686 on Build GRCh38
rs185873252 Chr.17: 81917730-81917730 on Build GRCh38
rs751456614 Chr.17: 81917788-81917788 on Build GRCh38
rs573315714 Chr.17: 81917761-81917761 on Build GRCh38
rs868815773 Chr.17: 81917662-81917662 on Build GRCh38

View Full Product Details